Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SCH 28080: A Potent Inhibitor of Gastric H+, K+–ATPase
Cell Signaling & Neuroscience
Prod. No. S 4443
Parietal cells (gastric acid (HCl) secreting cells) are present in the stomach and ilium and regulate the secretion of highly concentrated HCl into the lumen. They also recruit and recycle the transport protein H+,K+-ATPase, the primary gastric proton pump. An overactive pump may lead to ulcers and other problems; therefore, inhibiting an overactive pump is a major target for researchers. Several compounds have been shown to effectively inhibit this pump, including omeprazole (Prod. No. O-104) and the histamine H2 receptor antagonists cimetidine (Prod. No. C4522), ranitidine (Prod. No. R-101), famotidine (Prod. No. F6889) and nizatidine (Prod. No. N7035). Omeprazole, however, binds irreversibly, thus blocking acid secretion, leading to anacidity, hyperplasia and hypergastremia1.
Sigma-RBI now offers SCH 28080 (Prod. No. S4443), a potent, reversible inhibitor of gastric H+,K+-ATPase, which competitively binds to the luminal K+ high affinity site of H+,K+-ATPase. SCH-28080 does not inhibit Na+,K+-ATPase. In addition, SCH-28080 inhibits renal ouabain-insensitive H+,K+-ATPase, but not colonic ouabain-sensitive H+, K+-ATPase.2 The inhibition of ATPase activity by ouabain (Prod. No. O3125) has been widely used as a marker of Na+ pump activity in vitro, while inhibition by SCH-28080 has been used as a marker of H+-K+-ATPase activity3, as the binding sites for these compounds have only been found on one or the other of the ATPases. The binding site of SCH 28080 is located in the gastric H+,K+-ATPase a-subunit, in the first extracellular loop between the M1 and M2 transmembrane segments2, although further details are still being elucidated. 4,5
SCH 28080 has been shown to possess both antisecretory and cytoprotective activities. The antisecretory ED50 values obtained in the pylorus-ligated rat were 3.7 mg/kg p.o. and 2.8 mg/kg i.p., which were 7 and 10 times more potent than cimetidine (Prod. No. C4522), respectively. In rats, the compound's cytoprotective activity was demonstrated by inhibition of ethanol-induced gastric lesions in a dose-dependent manner (ED50: 3.0 mg/kg p.o.)6. In addition, SCH 28080 (1-30 mg/kg p.o. in rats) inhibited gastric ulcers provoked by aspirin, aspirin plus acid, indomethacin and stress6.
Clearly, SCH 28080 will continue to be an indispensable tool for the study of gastric secretion and the X+,K+-ATPases.
References
  1. Shamburek, R.D. and Schubert, M.L., Gastrenterol. Clin. North Am., 21, 527-550 (1992).
  2. Asano, S., et al., J. Biol. Chem., 272, 17668-17674 (1997).
  3. Codina, J., et al., Am. J. Physiol. Cell Physiol., 279, C1319-C1326 (2000).
  4. Farley, R.A., et al., J. Biol. Chem., 276, 2608-2615 (2001).
  5. Asano, S., et al., J. Biol. Chem., Published online ahead of printing Dec 29, 2003, www.jbc.org.
  6. Long, J.F., et al., J. Pharmacol. Exp. Ther., 114-20 (1983).
back to Cell Signaling