Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Product Lines
Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SR2640
Cell Signaling & Neuroscience
Selective CysLT1 leukotriene receptor antagonist.
Prod. No. S 7690
Leukotrienes exist as two distinct classes, hydroxyacids such as LTB4 (Prod. No. L 0517), and cysteinyl leukotrienes such as LTC4 (Prod. No. L 4886), LTD4 (Prod. No. L 5011) and LTE4. Recently cloned leukotriene receptors BLT1, CysLT1 and CysLT2 are members of the G protein-coupled receptor superfamily. Cysteinyl leukotrienes are important mediators of inflammatory responses, such as constriction of airways and vascular smooth muscle, increased endothelial membrane permeability leading to edema, and an enhanced secretion of viscous mucus. Their receptors have been implicated in a range of inflammatory diseases, specifically bronchial asthma. SR2640 acts as a selective CysLT1 leukotriene receptor antagonist.
  • Inhibits in a concentration-dependent manner the binding of 0.4 nM [3H]LTD4 to guinea pig lung membrane with an IC50 value of 23 nM resulting in inhibition of bronchoconstriction.1
  • Inhibits LTD4-stimulated increases in intracellular calcium and production of inositol phosphates in human polymorphonuclear neutrophils (PMN).3
  • In an in vivo model of chronic inflammatory bowel disease, SR2640 reduces inhibitory effects of LTD4 on LTB4-directed chemotaxis of PMN neutrophils by acting as systemic LTD4 receptor antagonist.4
Structure of S7690
References:
  1. Ahnfelt-Ronne, I., et al., Eur. J. Pharmacol., 155, 117-128 (1988).
  2. Bouchelouche, P.N., et al., Agents and Actions, 29, 1990, 299-307 (1990).
  3. Nielsen, O.H., et al., Prostaglandins Leukot. Essent. Fatty Acids, 42, 181-184 (1991).
back to Cell Signaling