Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SR 59230A
Cell Signaling & Neuroscience
The First Commercially Available Selective B3-Adrenoceptor Antagonist
Prod. No. S 8688
B-Adrenoceptors belong to the family of G protein-coupled receptors. They are widely distributed throughout central and peripheral nervous systems and in peripheral tissues. The three subtypes of these receptors have been identified, B1, B2, and B3. The B3 receptor subtype is the predominant type found in adipose tissue. Activation of these receptors stimulates lipolysis in white adipose tissue while increasing energy expenditure in brown adipose tissue.
  • Inhibits B3 adrenoceptor agonist-induced inhibition of spontaneous motility in the rat isolated colon, with a pA2 of 8.761
  • Is 100 times less potent as an antagonist of B1 or B2 mediated responses to isoproterenol (I 5627) or salbutamol (S 5013) in guinea pig tissues2
  • Blocks stimulation of cylic AMP synthesis in rat brown adipose tissue3
  • Inhibits B3 adrenoceptor-mediated relaxation in the human colon4
  • Reduces the inhibition of colonic motility and the thermogenic response of brown adipose tissue elicited in rats by BRL 37344 (a B3 selective receptor agonist, B-169) at oral doses2
References
  1. Manara, L. et al., Br. J. Pharmacol., 117, 435-442 (1996).
  2. Arch, J.R. and Kaumann, A.J., Med. Res. Rev., 13, 663-729 (1993).
  3. Nisoli, E., et al., Mol. Pharmacol., 49, 7-14 (1996).
  4. De Ponti, F., et al., Br. J. Pharmacol., 117, 1374-1376 (1996).
Click here to
    order