Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Tools for Phosphorylation Research
Cell Signaling & Neuroscience
p38 MAPK and Phospho-p38 MAPK (pThr180/pTyr182) ELISAS
P38 MAP kinase (MAPK, Mitogen-activated protein kinase), also known as a CDC-2-related protein kinase or CSBP (cytokine suppressive anti-inflammatory drug binding protein) 1 regulates many cellular processes including inflammation, cell differentiation, cell growth and death. p38 MAPK is activated in response to a variety of extracellular stimuli including osmotic shock, cytokines, LPS (Prod. No. L 2654) and anisomycin (Prod. No. A 9789). The activation of p38 MAPK is mediated by several upstream kinases that phosphorylate p38 at threonine 180 and tyrosine 182. This dually phosphorylated form of p38 is a High Osmolarity Glycerol response kinase (HOG), a 43 kDa endogenous, active form of p38 a, b, and g required for growth under high osmolarity conditions. This form is present in the variety of cells following exposure to a broad range of extracellular stimuli, including UV B irradiation of human skin, endotoxins, osmotic and environmental stress, and, most recently, during ischemia/reperfusion.2-5
Solid phase p38 MAPK ELISA (Prod. No. PM0100) and Phospho-p38 MAPK (pThr180/pTyr182) ELISA (Prod. No. CS0020) are designed for the in vitro quantitative detection of non-phopshorylated and dually phosphorylated human and mouse p38 MAPK in cell lysates. A monoclonal capture antibody specific for p38 MAPK (regardless of phosphorylation state) has been coated onto a 96 multiwell plate. p38 MAPK antigen in samples and standards binds to the capture antibody. A detection antibody specific for the phosphorylated or non-phosphorylated p38 MAPK is added and binds to the immobilized p38 MAPK protein. An anti-rabbit IgG-HRP completes the four-member sandwich. The reaction is visualized by a TMB substrate and the intensity of the color is directly proportional to the concentration of p38 MAPK present in the original sample. The reaction is read at 450 nm in a multiwell plate reader. The assays detect both native and recombinant p38 MAPK and phospho-p38 MAPK (pThr180/pTyr182) proteins. The assays are highly sensitive (below 16 pg/mL and below 0.8 units/mL, respectively), specific, fast (4 hours total time) and simple (offer precoated plates, all incubations proceed at room temperature and the reagents are supplied ready to use).
Size:   96 wells
Performance Characteristics:
  • Precision
       • Low intra-assay variability from 3.9% to 7.3%
       • Low inter-assay variability from 5.8% to 9.7%
  • Linearity of dilution – correlation coefficient for various dilutions was 0.99 for both assays
  • Recovery – average recovery was 93-97%
Storage:   Store at 2 to 8 °C
Components:
  • Standards: p38 MAPK (Prod. No. P 8870) and Phospho-p38 MAPK (pThr180/pTyr182) (Prod. No. P 7995) - lyophilized recombinant p38 MAPK and recombinant p38 MAPK dually phosphorylated by MKK6.
  • Standard/Sample diluent buffer
  • Monoclonal anti-p38 MAPK-coated 96 multiwell plate
  • Anti-p38 MAPK (Prod. No. P 7620) or Anti-phospho-p38 MAPK (pThr180/pTyr182) (Prod. No. P 7870)
  • Anti-rabbit IgG-HRP concentrate 100X
  • HRP Diluent
  • Wash Buffer Concentrate 25X
  • Stabilized TMB Chromogen
  • Stop Solution
  • Plate Sealers
References:
  1. Han, J., et al, Biochem. Biophys. Acta., 1265, 224-227 (2000).
  2. Bulavin, D.V., et al., Nature, 411, 102-107 (2001).
  3. Pfundt, R., et al., J. Pathol., 193, 248-255 (2001).
  4. Weinbrenner, C., et al., J. Mol. Cell Cardiol., 29, 2383-2391 (1997).
  5. Ge, B., et al., Science, 295, 1291-1294 (2002).
back to Cell Signaling