Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Product Lines
Apoptosis & Cell Cycle
Calcium Signaling
Cytokines & Growth
Cytoskeleton & ECM
Drug Metabolism
G Proteins
Gene Reg & Expression
Hormones
Immune CS & Blood
Ion Channels
Lipids in Cell Signaling
Neurobiology
Neurotransmission
Nitric Oxide & Cell Stress
Protein Phosphorylation
Signal Transduction
Product Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Nutrition Research
Obesity Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Quarterly
PathFinder
 Multi-Drug Resistance and Drug Metabolism
Cell Signaling & Neuroscience
The expression of multi-drug resistance (MDR) is key in the development of therapeutic and chemotherapeutic drugs. Accessibility, solubility, clearance rate, and drug-drug interactions can mean the difference between a wonder drug and a lethal drug. The classical form of MDR hinges on P-glycoprotein (Pgp) at the cell membrane. Pgp acts as a drug efflux pump, and provides a method to rid cells of both toxic and therapeutic compounds. Sigma has a comprehensive line of substrates, enzymes, antibodies and other screening reagents.
Drug metabolizing enzymes (DMEs) are a diverse group of proteins produced in the liver that are responsible for metabolizing a vast number of xenobiotic compounds. These compounds include exogenous proteins such as steroids, pharmaceuticals drugs, and environmental pollutants. Cytochrome P450 enzymes (CYPs) are known for their ability to metabolize a large number of structurally diverse compounds. Over 150 cytochrome P450 substrates, metabolites, inducers and inhibitors are highlighted.
Ultrafine (UC) Catalog Product Cross-referenced listings
Browse our complete line of MDR and Drug Metabolism products
Technical Resources:
Review Article: Multi-Drug Resistance in Cancer - Role of ABC Transporter Proteins
Pathway Slides:
Mechanisms In Multi-drug Resistance: ABC-transporters and detoxification enzymes I hepatocytes
ABC-ATPases as Regulatory Transporters for NHRs: chemical sensors for endogenous metabolites and xenobiotics
Domain Organization of Bile Salt Transporter (BSEP)
Domain Organization of Multi-drug Related Protein (MRP1)
Helpful external links
Cytochrome P450 - University of Colorado Health Sciences Center
Biocatalysis / Biodegradation Database - University of Minnesota
Cytochrome P450 Homepage
Click on the column headings to sort products. To re-alphabetize, hit the page refresh button in your web browser.
Product NameUltraFine CodeProduct #
AAMUUC432UC432
Acetaminophen, SigmaUltra, ≥99.0%UC491A7085
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, ≥98%UC297A4571
1-AminobenzotriazoleUC293A3940
4-Aminophenyl sulfone, 97%UC457A74807
AMMC iodide, ≥99% (NMR)UC460A1352
4-Androstene-3,17-dione, ≥98%UC300A9630
Arachidonic acid sodium salt, from porcine liver, ~99% (capillary GC), powderUC471A8798
Benzydamine N-oxide hydrogen maleate UC440UC440
1-Benzylimidazole, 99%UC464116416
7-Benzyloxyquinoline, 99% (by 1H-NMR), solidUC458B5182
7-Benzyloxy-4-trifluoromethylcoumarin, 99% (by 1H-NMR), solidUC462B5057
Bergamottin, purum, ≥98.0% (HPLC)UC43701338
(±)-Bufuralol hydrochlorideUC168UC168
Cannabidiol solution, drug standard, 1.0 mg/mL±5% in methanolUC500C6395
Carboxytolbutamide UC260UC260
ChlorzoxazoneUC338C4397
Clofibrate, liquidUC465C6643
Coumarin, ≥99% (HPLC)UC336C4261
3-Cyano-7-ethoxycoumarin, ≥97% (HPLC)UC455UC455
3-Cyano-7-methoxycoumarin UC454UC454
3-Cyanoumbelliferone, BioChemika, for fluorescence, ≥98.0% (TLC)UC49428605
Debrisoquine sulfateUC281D1306
O-Desmethylnaproxen UC305UC305
O-Desmethyltramadol UC304UC304
Dextromethorphan hydrobromide, meets USP testing specificationsUC337D9684
DextrorphanUC205UC205
Diclofenac sodium saltUC291D6899
3-[2-(Diethylamino)ethyl]-7-hydroxy-4-methylcoumarin hydrochloride, UC461188611
6´-7´-Dihydroxybergamottin UC441UC441
5,5-Diphenylhydantoin, ~99%UC295D4007
Dodecanoic acid, ≥99% (GC/titration)UC340L4250
Estrone 3-(β-D-glucuronide) sodium saltUC332E1752
7-Ethoxy-4-(trifluoromethyl)coumarin, ≥98% (TLC)UC449T2803
Furafylline, solidUC165F124
(±)-d5-Geosmin UC70UC70
Haloperidol metabolite IIUC302H102
Haloperidol, powderUC472H1512
Hydrocortisone, ≥98% (HPLC)UC425H4001
Hydrocortisone, meets USP testing specificationsUC425H3160
7-Hydroxy-4-(trifluoromethyl)coumarin, 98%UC299368512
Hydroxybufuralol maleate, ≥98% (HPLC)UC169UC169
6-Hydroxychlorzoxazone, ≥98% (HPLC)UC148UC148
6β-Hydroxycortisol, ≥98%UC426H6904
7-Hydroxycoumarin glucuronide sodium salt UC263UC263
7-Hydroxycoumarin sulfate potassium saltUC283UC283
(±)-4-Hydroxydebrisoquin sulfate UC261UC261
4′-Hydroxydiclofenac, Standard, ≥97% (HPLC)UC997H3661
12-Hydroxydodecanoic acid, 97%UC409198781
4-Hydroxymephenytoin UC126UC126
1´-Hydroxymidazolam UC430UC430
4-Hydroxymidazolam UC431UC431
(+)-3-Hydroxymorphinan hydrobromideUC497H156
(±)-4´-HydroxynirvanolUC134UC134
3′-p-Hydroxypaclitaxel, 95% (by 1H-NMR), solidUC998H3786
6α-HydroxytestosteroneUC286H3525
6β-HydroxytestosteroneUC287UC287
16α-Hydroxytestosterone  UC288UC288
16β-Hydroxytestosterone  UC289UC289
7α-Hydroxytestosterone  UC498UC498
2α-Hydroxytestosterone, ≥97%UC284H2773
2β-Hydroxytestosterone, ≥98%UC285H9526
6β-Hydroxytestosterone, ≥99% (TLC)UC282UC282
4-Hydroxytolbutamide, ≥98% (HPLC)UC160UC160
10-Hydroxywarfarin UC223UC223
4-Hydroxywarfarin UC209UC209
6-Hydroxywarfarin UC208UC208
7-Hydroxywarfarin UC211UC211
8-Hydroxywarfarin UC210UC210
Itraconazole, ≥98% (TLC)UC501I6657
Ketoconazole UC280UC280
(±)-MephenytoinUC177UC177
(R)-(−)-MephenytoinUC176UC176
(S)-(+)-MephenytoinUC175UC175
(±)-4´-MethoxymephenytoinUC131UC131
3-Methoxymorphinan hydrochlorideUC204M187
8-Methoxypsoralen, ≥98% (GC), powderUC466M3501
2-Methyl-1,2-di-3-pyridyl-1-propanone, 96%UC468856525
Midazolam hydrochloride UC429UC429
Midazolam-(methyl,1,3a,4-13C4-5-15N) maleate salt, ≥95% (excluding maleic acid, HPLC)UC601UC601
Monoacetyl dapsone UC428UC428
Morphine 3-β-D-glucuronideUC172M6886
Morphine 6-β-D-glucuronide hydrate, ≥90% (HPLC)UC17M3528
Morphine-3,6-diglucuronide tetrahydrateUC161UC161
Naloxone-3-glucuronide UC322UC322
Naloxone-3-glucuronide UC322UC322
Nifedipine, ≥98% (TLC), powderUC182N7634
(±)-NirvanolUC178UC178
(R)-(−)-NirvanolUC179UC179
(S)-(+)-NirvanolUC180UC180
Oleandomycin triacetateUC356T6514
Omeprazole, solidUC499O104
Oxidized NifedipineUC167UC167
Paclitaxel, from semisynthetic (from Taxus sp.), ≥97%UC290T7191
Paracetamol sulfate potassium salt UC448UC448
Phenacetin, powderUC492A2500
trans-2-Phenylcyclopropylamine hydrochlorideUC496P8511
Phenyl-d5-7-hydroxywarfarin UC602UC602
Proadifen hydrochloride, ≥95%UC292P1061
Progesterone 11alpha-glucuronideUC184UC184
Progesterone, ≥99%UC470P0130
Progesterone, meets USP testing specificationsUC470P3972
Quercetin dihydrate, ≥98% (HPLC), powderUC469Q0125
Quinidine, anhydrousUC298Q3625
Resorufin benzyl etherUC453B1532
Resorufin ethyl etherUC335E3763
Resorufin methyl etherUC330M1544
Resorufin pentyl etherUC493P0928
Resorufin sodium salt UC358R3257
Salicyl glucuronide disodium salt UC407UC407
Salicyl sulfate disodium salt UC301UC301
Sodium diethyldithiocarbamate trihydrate, puriss. p.a., for the determination and separation of heavy metals, ACS reagent, ≥99.0%UC35771480
Sulfaphenazole, ≥98% (HPLC)UC166UC166
Terfenadine UC199T9652
Terfenadine alcohol metaboliteUC206UC206
Testosterone, ≥98%UC339T1500
Tolbutamide UC294T0891
Tramadol hydrochlorideUC456UC456
(R)-(+)-Warfarin, ≥97% (HPLC)UC213UC213
(S)-(−)-Warfarin, ≥97% (HPLC)UC214UC214
back to top
back to Cell Signaling