|
Panorama® Antibody (Ab) Microarray provides a comprehensive system to establish protein expression profiles in biological samples. Based on a state-of-the-art manufacturing process, the antibody microarray contains 224 human antibodies to key cellular proteins with a special emphasis on cell signaling proteins. The antibodies are spotted in duplicate on a nitrocellulose-coated glass slide that has been blocked with a proprietary solution to eliminate background. The Panorama® Antibody Microarray reacts with samples from human, mouse, and rat. Additionally, the antibodies are also available separately for more extensive analysis of your sample.
- Accurate & robust protein expression profiling of your sample in less than half a day.
- Establish relative abundance of hundreds of proteins in less than 5 hours
- Suitable for use with Cy3/Cy5 –based detection chemistries
- Superior spot morphology with less than 10% variation
- Compatible with most commercially available scanners
Antibody List
Application Article
ScanArray® Express Slide Analysis Software (11 Kb TXT file)
GenePix® Pro Slide Analysis Software (18 Kb TXT file)
Technical Poster (222 kb PDF)
Brochure (298 kb PDF)
View Recent Literature
Slide Analysis Software (5 kb zip)
Order Information
| Product Code |
Product Description |
Technical Bulletin |
|
CSAA1 |
Panorama® Antibody Microarray — Cell Signaling Kit |
|
 |
Kit Contents:
Panorama® Antibody Slides — Cell Signaling (2 Each)
QuadriPERM Cell Culture Vessel (2 Each)
Extraction/Labeling Buffer
Protease Inhibitor Cocktail
Phosphatase Inhibitor Cocktail II
Benzonase®, Ultrapure
Array Incubation Buffer
Phosphate Buffered Saline, pH 7.4, with Tween 20 (Washing Buffer)
SigmaSpin Post-Reaction Clean-Up Columns
Collection Tubes, Polypropylene, 2 ml
Panorama® Antibody List — Cell Signaling |
ScanArray® Express is a registered trademark of PerkinElmer.
GenePix® Pro is a registered trademark of Molecular Devices.
QuadiPERM is a trademark of WC Heraeus GmbH, Ltd.
Tween is a registered trademark of the ICI Group.
Benzonase is a trademark of Merck KGaA.
Cy3 is a trademark of GE Healthcare.
Panorama is a registered trademark of Sigma-Aldrich.
Antibody Arrays
Arrays
Proteomics
Antibody Explorer
|