Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Mechanisms In Multi-Drug Resistance: ABC-transporters and detoxification enzymes I hepatocytes
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Mechanisms In Multi-Drug Resistance: ABC-transporters and detoxification enzymes I hepatocytes
The cellular components responsible for the phenomenon of multi-drug resistance (MDR) in the structural context of hepatocytes include intrinsic transmembrane proteins, generically called drug efflux pumps, and the enzymes responsible for detoxification and biotransformation of xenobiotics. The enzymes include the cytochrome P450s (CYPs) and the phase II conjugation enzymes such as glutathione-S-transferase (GST). The major drug efflux pumps are in the superfamily of ABC (ATP-binding-cassette)-transporters. Of these ABC-ATPases, the major subfamilies are the MDR/TAPs and MRPs (multi-drug resistance related proteins). The natural substrates are indicated for the drug efflux pumps.
Back to Top
References:
Shabbits, J.A., et al., Molecular and pharmacological strategies to overcome Multidrug resistance. Expert Rev. Anticancer Ther., 1, 585-594 (2001).
Di, P.A., et al., Modulation by flavonoids of cell multidrug resistance mediated by P-glycoprotein and related ABC transporters. Cell. Mol. Life Sci., 59, 307-322 (2002).
Meier, P.J., and Stiger, B., Bile salt transporters. Annu. Rev. Physiol., 64, 635-661 (2002).