Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Caspase Cascade
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Caspase Cascade
Apoptosis or programmed cell death is triggered by a variety of stimuli, including cell surface receptors like FAS, the mitochondrial response to stress, and factors released from cytotoxic T cells. The caspases comprise a class of cysteine proteases many members of which are involved in apoptosis. The caspases convey the apoptotic signal in a proteolytic cascade, with caspases cleaving and activating other caspases that subsequently degrade cellular targets that lead to cell death. The activating caspases include caspase-8 and caspase-9. Caspase-8 is the initial caspase activated in response to receptors with a death domain that interacts with FADD. The mitochondrial stress pathway begins with the release of cytochrome c from mitochondria, which then interacts with Apaf-1, causing self-cleavage and activation of caspase-9. The effector caspases, caspase-3, -6 and-7 are downstream of the activator caspases and act to cleave various cellular targets. Granzyme B and perforin, proteins released by cytotoxic T cells, induce apoptosis in target cells by forming transmembrane pores and triggering apoptosis, perhaps through cleavage of caspases. Caspase independent mechanisms of granzyme B-mediated apoptosis have been suggested.
Back to Top
References:
Earnshaw, W.C., et al., Mammalian caspases: structure, activation, substrates, and functions during apoptosis. Ann. Rev. Biochem., 68, 383-424 (1999).
Adrain, C., and Martin, S.J., The mitochondrial apoptosome: a killer unleashed by the cytochrome seas. Trends Biochem. Sci., 26, 390-397 (2001).