Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Product Lines
Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Lipids in Cell Signaling
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Lipids in Cell Signaling
Many of the lipids involved as second messengers in cell signaling pathways arise from the arachidonic acid (AA) pathway. AA is an unsaturated fatty acid that is a normal constituent of membrane phospholipids and is released from the phospholipids by the actions of phospholipase A2 (PLA2). Prostaglandins (PG) arise from a cyclic endoperoxide generated by the enzyme system PG synthetase, a complex of enzymes that includes cyclooxygenase (COX). There is a constitutive (COX-1) and an inducible cyclooxygenase (COX-2). The cyclic endoperoxide intermediate is also a precursor of prostacyclin (PGI2) and thromboxane (TXA3). Other groups of compounds in this class, leukotrienes (LT) and lipoxins (LP), are derived directly from AA without the mediation of a cyclic endoperoxide. Lipoxygenase acts on AA to produce 5-hydroperoxyeicosatetraenoic acid (5-HPETE) that is converted to LTA4. LTA4 is the precursor of LTB4, that induces inflammation by its chemotactic and degranulating actions on polymorphonuclear lymphocytes (PML), and of LTC4, LTD4, and LTE4, the amino acid containing LTs that induce vasoconstriction and bronchoconstriction and are involved in asthma and anaphylaxis.
Back to Top
References:
Heller, A., et al., Lipid mediators in inflammatory disorders. Drugs, 55, 487-496(1998).
Cook, J.A., et al., Prostaglandins, thromboxanes, leukotrienes, and cytochrome P-450 metabolites of arachidonic acid. New Horiz., 1, 60-69(1993).
Dubois, R.N., et al., Cyclooxygenase in biology and disease. FASEB J., 12, 1063-1073(1998).