Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Diversity of G Protein-Coupled Receptor Signal
Transduction Pathways
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Diversity of G Protein-Coupled Receptor Signal
Transduction Pathways
Receptors coupled to heterotrimeric GTP-binding proteins (G proteins) are integral transmembrane proteins that transduce extracellular signals to the cell interior. G protein-coupled receptors exhibit a common structural motif consisting of seven membrane spanning regions. Receptor occupation promotes interaction between the receptor and the G protein on the interior surface of the membrane. This induces an exchange of GDP for GTP on the G protein a subunit and dissociation of the a subunit from the bg heterodimer. Depending on its isoform, the GTP-a subunit complex mediates intracellular signaling either indirectly by acting on effector molecules such as adenylyl cyclase (AC) or phospholipase C (PLC), or directly by regulating ion channel or kinase function.
Back to Top
References:
Schoneberg, T., et al., Structural basis of G protein-coupled receptor function. Mol. Cell. Endocrinol., 151, 181-193 (1999).
LeVine, H., 3rd., Structural features of heterotrimeric G protein-coupled receptors and their modulatory proteins. Mol. Neurobiol., 19, 111-149 (1999).
Morris, A.J., et al., Physiological regulation of G protein-linked signaling. Physiol. Rev., 79, 1373-1430 (1999).