Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Fas and Related Proteins with Death Domains
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Fas and Related Proteins with Death Domains
The death domain (DD) of Fas (FADD) shows high-sequence homology to the Drosophila melanogaster protein reaper. The death domain is also found in tumor necrosis factor receptor 1 (TNF-R1), TNF-R1-associated death domain (TRADD), Fas-associated death domain (FADD), and Fas receptor-interaction protein (RIP). Fas and FADD associate through their homologous DD while TNF-R1 and TRADD associate through their homologous DD.
Back to Top
References:
Baker, S.J., and Reddy, E.P., Modulation of life and death by the TNF receptor superfamily. Oncogene., 17, 3261-3270 (1998).
Stanger, B.Z., Looking beneath the surface: the cell death pathway of Fas/APO-1 (CD95). Mol. Med., 2, 7-20 (1996).
Golstein, P., et al., Homology between reaper and the cell death domains of Fas and TNF-R1. Cell, 81, 185-186 (1995).