Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Product Lines
Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Inositol Phosphates
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Inositol Phosphates
Inositol phosphates are important intracellular second messengers in eukaryotic cells. Inositol, a cyclic molecule with six hydroxyl groups, forms the hydrophobic head group of membrane inositol phospholipids. The actions of lipid kinases and phosphatases on the inositol ring of inositol phospholipids generate a variety of phosphoinositol phospholipids. As a result of upstream signaling events, specific phospholipases are activated, cleave phosphoinositol phospholipids, and release intracellular inositol phosphate second messenger molecules. The most studied inositol phosphate second messenger is inositol 1,4,5-trisphosphate (IP3). IP3 is released from phosphatidylinositol 4,5-bisphosphate (PIP2) by the action of phospholipase C-b and is responsible for the release of calcium ions from intracellular stores in the endoplasmic reticulum.
Back to Top
References:
Irvine, R.F., and Schell, M.J., Back in the water: the return of inositol phosphates. Nat. Rev. Mol. Cell Biol., 2, 327-338 (2001).
Shears, S.B., Assessing the omnipotence of inositol hexakisphosphate. Cell. Signal., 13, 151-158 (2001).