Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Insulin Pathway
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Insulin Pathway
Signaling through the insulin pathway is critical for the regulation of intracellular and blood glucose levels and the avoidance of diabetes. Insulin binds to its receptor leading to the autophosphorylation of the b-subunits and the tyrosine phosphorylation of insulin receptor substrates (IRS). IRS phosphorylates the SH2 domain of Shp2, a tyrosine phosphatase, and the SH3 domain of the adaptor molecule Grb2. Activated Grb2 recruits Sos1 that, in turn, activates the Ras signaling pathway and gene transcription. IRS also activates phosphoinositide 3-kinase (PI3K) through its SH2 domain, thus increasing the intracellular concentration of PIP2 and PIP. This, in turn, activates phosphatidylinositol phosphate-dependent kinase-1 (PDK-1), that subsequently activates Akt/PKB This results in the translocation of the glucose transporter (GLUT4) from cytoplasmic vesicles to the cell membrane.
Back to Top
References:
Bevan, P., Insulin signalling. J. Cell Sci., 114, 1429-1430 (2001).
Kido, Y., et al., Clinical review 125: The insulin receptor and its cellular targets. J. Clin. Endocrinol. Metab., 86, 972-979 (2001).