Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Product Lines
Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Nitric Oxide Metabolism
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Nitric Oxide Metabolism
Nitric oxide (NO) is as a major signaling molecule in neurons and in the immune system, either acting within the cell in which it is produced or by penetrating cell membranes to affect adjacent cells. Nitric oxide is generated from arginine by the action of nitric oxide synthase (NOS). NO has a half-life of only a few seconds in vivo . However, since it is soluble in both aqueous and lipid media, it readily diffuses through the cytoplasm and plasma membranes. NO has effects on neuronal transmission as well as on synaptic plasticity in the central nervous system. In the vasculature, NO reacts with iron in the active site of the enzyme guanylyl cyclase (GC), stimulating it to produce the intracellular mediator cyclic GMP (cGMP), that in turn enhances the release of neurotransmitters resulting in smooth muscle relaxation and vasodilation. NO may also be involved in the regulation of protein activity through S-nitrosylation. In the extracellular milieu, NO reacts with oxygen and water to form nitrates and nitrites. NO toxicity is linked to its ability to combine with superoxide anions (O2-) to form peroxynitrite (ONOO-), an oxidizing free radical that can cause DNA fragmentation and lipid oxidation. In the mitochondria, ONOO- acts on the respiratory chain (I-IV) complex and manganese superoxide dismutase (MnSOD), to generate superoxide anions and hydrogen peroxide (H2O2), respectively.
Back to Top
References:
Brown, G.C., Nitric oxide and mitochondrial respiration. Biochim. Biophys. Acta., 1411, 351-369 (1999).
Lipton, S.A., Neuronal protection and destruction by NO. Cell Death Differ., 6, 943-951 (1999).
Murad, F., Nitric oxide signaling: would you believe that a simple free radical could be a second messenger, autacoid, paracrine substance, neurotransmitter, and hormone? Recent Prog. Horm. Res., 53, 43-60 (1998).