|
Schematic Representation of GABAB Receptor
Representation of the GABAB receptor heterodimer which is composed of two, seven transmembrane-spanning units that are linked by their carboxyl-termini. The apparent ligand binding domains are on the extracellular surface of R1 and R2. Activation of the heterodimer modulates adenyl cyclase
activity via G-proteins that, in addition, influence K+ and Ca2+ conductance.
Back to Top
References
Marshall, F.H., et al., GABAB receptors - the first 7TM heterodimers. Trends Pharmacol. Sci., 20, 396-399 (1999).
Couve, A., et al., GABAB receptors : a new paradigm in G protein signaling. Mol. Cell. Neurosci., 16, 296-312 (2000).
|