Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
The p53 Signaling Pathway
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
The p53 Signaling Pathway
The tumor-suppressor protein p53 exhibits sequence-specific DNA-binding, directly interacts with various cellular and viral proteins, and induces cell cycle arrest in response to DNA damage. In response to signals generated by a variety of genotoxic stresses, e.g, UV irradiation or DNA damage, p53 is expressed and undergoes post-translational modification that results in its accumulation in the nucleus. The p53-dependent pathways help to maintain genomic stability by eliminating damaged cells either by arresting them permanently or through apoptosis. For example, g-irradiation activates p53 to turn on the transcription of p21CIP1, which, in turn, binds to and inhibits cyclin-dependent kinases, causing hypophosphorylation of retinoblastoma (Rb), thus preventing the release of E2F and blocking the G1-S transition. Some of the cellular effects of p53 can be blocked by the deregulated expression of c-Myc, Bcl-2, or E2F. p53 activity is controlled through an autoregulatory loop involving Mdm2. The binding of Mdm2 to p53 targets p53 for degradation and inhibits p53-induced cell-cycle arrest and apoptosis.
Back to Top
References:
Gu, J., et al., Identification of a sequence element from p53 that signals for Mdm2-targeted degradation. Mol. Cell Biol., 20, 1243-1253 (2000).
Jimenez, G.S., et al., p53 regulation by post-translational modification and nuclear retention in response to diverse stresses. Oncogene, 18, 7656-7665 (1999).
King, K.L., and Cidlowski, J.A., Cell cycle regulation and apoptosis. Annu. Rev. Physiol., 60, 601-617 (1998).