Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
VEGF Receptor Signaling
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
VEGF Receptor Signaling
Vascular endothelial growth factor (VEGF) stimulates mitogenesis of endothelial cells and increases vessel permeability. VEGF exerts its effects by binding to one of its three receptors (VEGFR-1/Flt-1, VEGFR-2/KDR/Flk-1, VEGFR-3/Flt-4). Upon binding of the ligand the receptor under goes transautophosphorylation and subsequent activation of a cell type-dependent signaling cascade. Signaling cascades that have been implicated in VEGF action include Src, PLC-g, MAPK, as well as STAT3 and STAT5. VEGF initiates embryonic vasculogenesis and triggers angiogenic sprouting via its activation of VEGFR-2 on vascular endothelial cells. VEGF activation of VEGFR-1 may limit cell proliferation induced by activation of VEGFR-2 and, thus, may limit VEGF-induced angiogenesis.
Back to Top
References:
Yancopoulos, G.D., et al., Vascular-specific growth factors and blood vessel formation. Nature, 407, 242-248 (2000).
Clauss, M., Molecular biology of the VEGF and the VEGF receptor family. Semin. Thromb. Hemost., 26, 561-569 (2000).
Neufeld, G., et al., Vascular endothelial growth factor (VEGF) and its receptors. FASEB J., 13, 9-22 (1999).