|
Voltage-Gated Calcium Channels
Voltage-gated calcium channels are heteromultimers composed of an a1 subunit and three auxiliary subunits, a2-d, b and g. The a1 subunit forms the ion pore and possesses gating functions and, in some cases, drug binding sites. Ten a1 subunits have been identified, which, in turn, are associated with the activities of the six classes of calcium channels. L-type channels have a1C (cardiac), a1D
(neuronal/endocrine), a1S (skeletal muscle), and a1F (retinal) subunits; N-type channels have a1B subunits; P- and Q-type channels have a1A subunits, and T-type channels have a1G, a1H, and a1I subunits. The a1 subunits each have four homologous domains (I-IV) that are composed of six transmembrane helices. The fourth transmembrane helix of each domain contains the voltage-sensing function. The four a1 domains cluster in the membrane to form the ion pore. The b-subunit is localized intracellularly and is involved in the membrane trafficking of a1 subunits. The
g-subunit is a glycoprotein having four transmembrane segments. The a2 subunit is a highly glycosylated extracellular protein that is attached to the membrane-spanning d-subunit by means of disulfide bonds. The a2-domain provides structural support required for channel stimulation, while the d domain modulates the voltage-dependent activation and steady-state inactivation of the channel.
Back to Top
References:
Jones, S.W., Overview of voltage-dependent calcium channels. J. Bioenerg. Biomembr., 30, 299-312 (1998).
|