Epigenetics
Bisulfite DNA Modification
ChIP
DNA Methylation Detection
DNA Purification
Key Resources
Post Reaction
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Sigma has a wide portfolio of products to support Epigenetics research. The Imprint™ Bisulfite Modification Kit is a rapid method to determine the methylation status of CpG islands. The Imprint Chromatin Immunoprecipitation Kit provides the perfect blend of speed and performance for the investigation of protein-DNA interactions in vivo. Sigma also has products to study Histone modification, including transcription regulators such as anti-histone antibodies.
Tool Box
  • Your Favorite Gene Search
  • Learning Center
  • Publications
  • Contact Us
  • Epigenetics Overview
  • Technologies of Interest
  • Protocols
  • Promotions
  • US Promotions
  • Products
  • Online Product Catalog
  • DNA Purification
  • Imprint DNA Modification Kit
  • Imprint ChIP Kit
  • Genetics Books
  • Labware
  • Related Topics
  • Research Essentials
  • Functional Genomics
  • Gene Regulation
  • The Imprint Chromatin Immunoprecipitation Kit
    The fastest and most convenient ChIP kit available
    • Fast - Total protocol time of less than 6 hours, making the Imprint kit the fastest on the marketing
    • Sensitive - As few as 20,000 cells required for each ChIP sample
    • Convenient - fewest steps of any available ChIP protocol
    • Flexible - Protocols for cells or tissue, and convenient strip-well format for high throughput applications.
    • Complete - includes columns and reagents for DNA purification as well as an integrated protocol for amplification with the GenomePlex technology
    Image of Antibodies sign up icon
    Prestige Antibodies
    Search for RNAi reagents, antibodies, bioactive small molecules and proteins