Epigenetics
Bisulfite DNA Modification
ChIP
DNA Methylation Detection
DNA Purification
Key Resources
Post Reaction
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Imprint™ Chromatin Immunoprecipitation Kit
Epigenetics
The Fastest and Most Convenient ChIP Kit Available! Header
Sigma® combines speed and convenience with performance in the Imprint™ Chromatin Immunoprecipitation Kit (CHP1). The Imprint ChIP Kit provides a complete solution for Chromatin Immunoprecipitation including columns and reagents for DNA purification and an integrated protocol for ChIP DNA amplification with our GenomePlex® Whole Genome Amplification Kit (WGA2). The flexible format allows for immunoprecipitation and purification of DNA from mammalian cells or tissue in a convenient strip-well format.
Did you know... Graphic
  • Fast—Total protocol time of less than 6 hours making the Imprint kit the fastest on the market
  • Sensitive—As few as 20,000 cells required for each ChIP sample
  • Convenient—Fewest steps of any available ChIP protocol
  • Flexible—Protocols for cells or tissue, and convenient
    strip-well format for high-throughput applications
  • Complete—Includes columns and reagents for DNA purification as well as an integrated protocol for amplification with the GenomePlex technology
View the poster Rapid and easy method for Chromatin IP (ChIP) analysis of metastatic cells
(446 KB PDF).
Figure 1 - Protocol Time Graph Figure 1. Comparison of time required for protocol completion from fixation through purification using different Chromatin Immunoprecipitation kits. Each ChIP experiment was carried out using each manufacturers recommended protocol and total required times were compared.
Figure 2 - RNA Pol2 Antibody Percent Input Profile Graph
Figures 2 and 3. A highly sensitive ChIP assay. HeLa cells were counted, fixed, and immunoprecipitated according to the Imprint CHP1 bulletin with optional High Sensitivity method. ChIP assays were performed using anti-H3K9ac (H9286) and kit provided antibodies against: human RNA Polymerase II and non-specific mouse IgG. To evaluate ChIP enrichment, SYBR qPCR was conducted targeting the GAPDH promoter region (highly expressed house-keeping gene). Percent Input describes the amount of DNA with and without antibody selection. For these antibodies, apparent enrichment increased with fewer cells per reaction from diminished non-specific pull-down.
Figure 3 - H3K9ac Antibody Percent Input Profile Graph
Ordering Information
Product # Product Name
CHP1-24RXN
CHP1-96RXN
Imprint Chromatin Immunoprecipitation Kit
Related Products
Product # Product Name
T6789 Tris Buffered Saline, with BSA, pH 8.0
H0913-200UL Anti-Monoclonal Anti-acetyl-Histone H3 (Ac-Lys9) antibody produced in mouse