|
Sigma provides a variety of kits that may be used to isolate DNA to begin your Epigenetic workflow. Our GenElute product line provides a simple and convenient way to purify high-quality Genomic DNA from mammalian cells and tissues, blood, and plants. Each kit utilizes a spin column format to rapidly extract Genomic DNA free of contaminants. The purified product is ideal for use in DNA Methylation analysis.
GenElute Mammalian Genomic DNA Miniprep Kit |
DNA yields of 25-30 µg can be isolated from up to 2x106 cells or 25mg of tissue in less than 40 minutes. All reagents, columns and tubes provided.
G1N10 10 Purifications
G1N70 70 Purifications
G1N350 350 Purifications
For additional information visit the GenElute Mammalian DNA Miniprep Kit page
|
|
GenElute Blood Genomic DNA Kit |
DNA may be isolated from fresh or aged blood in less than 40 minutes. All contaminants are removed by provided prewash solution, an Rnase solution is provided as well.
NA2000 10 Purifications
NA2010 70 Purifications
NA2020 350 Purifications
For additional information visit the GenElute Blood Genomic DNA Kit page
|
|
GenElute Plant Genomic DNA Miniprep Kit |
Up to 100 mg fresh or 20 mg of freeze-dried plant tissue can result in DNA yields of 20 µg per prep. Individual preps take no longer than 40 minutes and do not require Rnase A digestion.
G2N10 10 Purifications
G2N70 70 Purifications
G2N350 350 Purifications
For additional information visit the GenElute Plant Genomic DNA Kit page
|
Epigenetics Overview
back to the Epigenetics homepage
|