|
Epigenetics is the study of heritable changes in gene expression without a change in DNA sequence. The best understood Epigenetic mechanism, DNA methylation, refers to the addition of a methyl group by the enzyme DNA Methyltransferase to the 5-carbon of cytosine in a CpG dinucleotide. Methylation in regulatory regions adjacent to genes generally acts to suppress gene expression and/or regulation potentially having an impact on cellular function. Once a cell has an established DNA Methylation pattern, methlyated sites are inherited by daughter cells which have important implications in cellular function. Epigenetic research has demonstrated that aberrant DNA methylation is present in several disease states including cancer, in addition to other genetic diseases. Carcinogenesis can occur when DNA methylation acts to silence tumor suppressor genes, leading to heritable alterations of these genes.
Imprint DNA Modification Kit
Bisulfite DNA Modification is a technique to analyze gene-specific DNA methylation that converts unmethylated cytosine to uracil while methylated cytosine remains unaltered. Methyl-Specific PCR and DNA sequencing may be performed downstream of Bisulfite Modification to elucidate Epigenetic patterns and their impact on cellular function. Sigma provides the Imprint DNA Modification Kit that rapidly and accurately modifies unmethylated cytosine resulting in samples that are ready for all downstream applications.
Epigenetic Workflow
Sigma has a portfolio of products for your Epigenetic studies including DNA Purification, Detection and Post Reaction analysis that includes Methylation-Specific PCR and Sequencing. Please click on the Epigenetic Workflow below to learn more about Sigma’s product solutions for your Epigenetic analyses.
back to the Epigenetics homepage
|