|
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on
BioSupplyNet.com. |
Selected Protocol from "Gene Transfer: Delivery and Expression of DNA and
RNA", Edited by Theodore Friedmann and John Rossi.
Genetic Manipulation of Mammalian Cells by
Microinjection
David W. Rose
Department of Medicine and Moores Cancer Center, University of California, San
Diego, La Jolla, California 92093-0673
ABSTRACT
Of the many methods available for the introduction of DNA and other biological
materials into cells, microinjection of individual mammalian cells is one of
the more technically difficult to perform. The technique is also reliant upon
comparatively expensive equipment and demands the experimental skills of an
experienced operator. There are, however, very good reasons to use this approach.
It allows the introduction of molecules into a defined population of cells at a
defined concentration, and the timing of the experiment can be controlled
stringently, minimizing problems associated with overexpression. Perhaps the most
powerful aspect of microinjection is the ability to introduce several types of
reagents into cells simultaneously, including DNA constructs, a labeled dextran to
mark the injected cells, antibodies, short interfering RNAs (siRNAs), and peptides
(Lavinsky et al. 1998; Rose et al. 1998; Jepsen et al. 2000; Zhu et al. 2006). No
other techniques available provide these capabilities.
Delivery by microinjection can be used for any type of cell that is adherent in
culture, including primary cells. Because siRNA can be easily and rapidly
generated for any target gene, it is relatively simple to assess many effects of
the knockout of any gene in any type of adherent cell in a matter of a few days.
In addition, delivery by microinjection assures that every cell receives the
siRNA at a relatively equal concentration. Many of the disadvantages of the
approach relate to the challenging aspects of the technique itself: It is not
trivial to learn, and it is an undertaking that requires attention to detail. It
also suffers the technical disadvantage that a limited number of cells are
involved, which often does not permit the subsequent analysis of effects upon some
biochemical parameters. The major limitation of the approach is the small amount
of material obtained, but as the sensitivity of analysis techniques increases, so
does the usefulness of this method.
Products Available for this Protocol
| Protocol Material Description |
Product # |
Product Name |
Add to Cart |
| Buffers,
Solutions, and Reagents |
|
|
|
| Dextran, fluorescently labeled |
46945 |
Fluorescein isothiocyanate–dextran |
|
| Oligonucleotide siRNAs |
siRNAs |
Oligonucleotide siRNAs |
|
| 5 mM Sodium phosphate |
342483 |
Sodium phosphate |
|
| KCl |
P9541 |
Potassium chloride |
|
| Glucose |
G5400 |
D-(+)-Glucose |
|
back to top
View all available Cold Spring Harbor Laboratory Press
Protocols
Product Association Disclaimer:
The Sigma-Aldrich products listed for this specific protocol were selected either
to match or to supplement the products listed within the actual protocol. The
products/reagents from Sigma-Aldrich have been qualified for usage, but may not
have been validated for this specific application. Please refer to the detailed
product description on the usage of specific products of interest.
|