|
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on
BioSupplyNet.com. |
Selected Protocol from "Gene Transfer: Delivery and Expression of DNA and RNA", Edited by Theodore
Friedmann and John Rossi.
Knockdown Transgenic Mice Generated by Silencing Lentiviral Vectors
Oded Singer, Gustavo Tiscornia, and Inder M. Verma
The Salk Institute for Biological Studies, Laboratory of Genetics, La Jolla, California 92037
ABSTRACT
This protocol describes the use of lentiviral vectors to deliver genes and small interfering RNA (siRNA)-expressing cassettes into preimplantation embryos. This technique allows for the rapid development of transgenic animals and transgenic knockdown animals.
For the purpose of generating transgenic and knockout animals, preimplantation embryos must be harvested and manipulated in vitro. Although the generation of transgenic animals has been performed by pronuclear injection of DNA into single-cell embryos, the generation of mouse knockouts is time-consuming and laborious. An embryonic stem (ES) knockout line must be generated, characterized, and injected into a blastocyst in order to obtain a chimeric founder that can be subsequently bred to homozygosity. Taking advantage of the unique ability of lentiviral vectors to generate transgenic animals (Lois et al. 2002; Pfeifer et al. 2002), lentiviruses expressing short hairpin RNAs (shRNAs) from polymerase III (pol III) promoters such as H1 (Tiscornia et al. 2003) and mU6 (Rubinson et al. 2003) can be used to generate transgenic knockdown mice. Two methods are available for delivering lentiviral particles to the embryo to obtain lentiviral transgenesis: zona pellucida removal and subzonal injection. For a detailed description of harvesting and manipulating mouse embryos, see Hogan et al. (1994). The zona pellucida removal protocol is able to achieve up to 100% transgenesis and does not require the use of a micromanipulator. However, zona removal is toxic to the embryos and results in low survival of embryos. The subzonal injection protocol is able to achieve up to 100% transgenesis and is not toxic for the embryos; therefore, survival of embryos is much higher. Nevertheless, the use of micromanipulators requires some practice.
Pellucida Removal and Subzonal Injection Methods
This protocol describes two methods to deliver genes and siRNA-expressing cassettes into preimplantation mouse embryos using lentiviral vectors.
Products Available for this Protocol
| Protocol Material Description |
Product # |
Product Name |
Add to Cart |
| Buffers, Solutions, and Reagents |
|
|
|
| H2O |
W4502 |
Water |
|
| NaCl |
S3014 |
Sodium chloride |
|
| KCl |
P9541 |
Potassium chloride |
|
| CaCl2•2H2O |
C7902 |
Calcium chloride dihydrate |
|
| MgCl2•6H2O |
M1028 |
Magnesium chloride solution |
|
| Glucose |
G5400 |
D-(+)-Glucose |
|
| Polyvinylpyrrolidone (PVP) |
P5288 |
Polyvinylpyrrolidone |
|
| Gonadotropin from pregnant mare serum |
G4527 |
Gonadotropin from pregnant mare serum |
|
| Human chorionic gonadotropin |
C8554 |
Chorionic gonadotropin human |
|
| Hyaluronidase solution |
H3884 |
Hyaluronidase from bovine testes |
|
| Mineral oil |
M5904 |
Mineral oil |
|
| Media |
|
|
|
| M2 |
M7167 |
M2 medium |
|
| M16 |
M7292 |
M16 medium |
|
back to top
View all available Cold Spring Harbor Laboratory Press Protocols
Product Association Disclaimer:
The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the
products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for
usage, but may not have been validated for this specific application. Please refer to the detailed product
description on the usage of specific products of interest.
|