Functional Genomics and RNAi
Synthetic RNA
MISSION What's New
Key resources
RNAi Learning Center
shRNA
siRNA
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 TargeTron® Gene Knockout System
Functional Genomics & RNAi
Genetic Engineering that is Right on Target!
Introduction to TargeTron
The TargeTron Gene Knockout System (TA0100) is a revolutionary method for rapid and specific disruption of genes in prokaryotic organisms. Utility of the technology has been demonstrated for prokaryotic genetic engineering, systems biology and functional genomics approaches.
View Technical Animation and NEW presentations on TargeTron and group II intron biology!
Features
  • Targeted and permanent gene disruption
  • Simple, streamlined protocol; Knockouts in 3 days or less
  • Minimal screening to isolate mutants
  • >90% successful targeted insertion
  • No specific host factor requirements
  • Pre-screen your target sequence on the TargeTron Test Site
Resources
TargeTron Vectors
Detailed Description and Algorithm
TargeTron Test Site
TargeTron Design Site
User Guide (1.1 MB PDF)
pACD4K-C Vector Sequence (7.7 KB TXT)
Updated License Agreement (86 KB PDF)
Poster (1.6 MB PDF)
Frequently Asked Questions
References
Ordering Information
Host Range
TargeTron has been validated in multiple organisms; however, work to adapt the system to other organisms is ongoing and data will be provided as it becomes available.
Organism Reference
Agrobacterium tumefaciens Gene Targeting in Gram-Negative Bacteria by Use of a Mobile Group II Intron ("Targetron") Expressed from a Broad-Host-Range Vector
Bacillus subtilis (TargeTron Vector pNL9164, Product No. T6701)
Clostridium acetobutylicum The ClosTron: A universal gene knock-out system for the genus Clostridium
Clostridium botulinum The ClosTron: A universal gene knock-out system for the genus Clostridium
Clostridium difficile The ClosTron: A universal gene knock-out system for the genus Clostridium
Clostridium perfringens (TargeTron Vector pJIR750ai, Product No. T7701) Disruption of a toxin gene by introduction of a foreign gene into the chromosome of Clostridium perfringens using targetron-induced mutagenesis
Clostridium sporogenes The ClosTron: A universal gene knock-out system for the genus Clostridium
Escherichia coli (TargeTron Gene Knockout System, Product No. TA0100)
Lactococcus lactis (TargeTron Gene Knockout System, Product No. TA0100)
Proteus mirabilis Reference in press
Pseudomonas aeruginosa Gene Targeting in Gram-Negative Bacteria by Use of a Mobile Group II Intron ("Targetron") Expressed from a Broad-Host-Range Vector
Salmonella typhimurium (TargeTron Gene Knockout System, Product No. TA0100)
Serratia marcescens Unpublished to date
Shigella flexneri (TargeTron Gene Knockout System, Product No. TA0100)
Staphylococcus aureus (TargeTron Vector pNL9164, Product No. T6701)
Xylella fastidiosa Unpublished to date
Back to Top
Related Products
Below is an easy guide to materials needed for or that complement the TargeTron Gene Knockout System.
Related Technologies
Functional Genomics
Sigma-Genosys
This product and its use are the subject of one or more of U.S. Patent Nos. 5,698,421, 5,804,418, 5,869,634, 6,027,895, 6,001,608, and 6,306,596 and/or other pending U.S. and foreign patent applications controlled by InGex, LLC.
TargeTron is a registered trademark of InGex, LLC.
Back to Top