Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Synthetic RNA
MISSION What's New
Learning Center
Zinc Finger Nuclease Technology
miRNA
shRNA
siRNA
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 TargeTron® Vector pJIR750ai
Functional Genomics & RNAi
TargeTron vector pJIR750ai is a circularized vector that can be used for targeted gene knockouts in gram-positive bacteria such as Clostridium perfringens type A.1 Expression of the group II intron RNA is under the control of the β-2 toxin gene promoter, cpb2.1 As supplied, the pJIR750ai vector is re-targeted to the α-toxin (plc) gene. α-Toxin, a phospholipase C, is an extracellular toxin produced by all C. perfringens isolates.1 For validation in a specific C. perfringens strain, this vector can be used directly to knockout plc without any further modifications to the enclosed protocol. In order to re-target this vector to knockout other genes, the plc specific IBS-EBS fragment (350 bp) between the Hind III and BsrG I sites can be cut out and replaced with another gene specific fragment. For re-targeting protocols, refer to the TargeTron Gene Knockout System, TA0100 User Guide (1.1 MB PDF).
Technical Bulletin
Vector Map
Ordering Information
References
  1. Chen, Y. et. al, Appl Environ Microbiol. 71(11): 7542-7547(2005).
Related Products
TargeTron Gene Knockout System
TargeTron Vector
pJIR750ai Vector Map
TargeTron is a registered trademark of InGex, LLC.
back to top
TargeTron® Gene Knockout System