Functional Genomics and RNAi
Synthetic RNA
MISSION What's New
Key resources
RNAi Learning Center
shRNA
shRNA Search & Order
Learning Center
Promotions
Products
Labware
Academic Programs
Related Topics
siRNA
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 MISSION® shRNA Product Overview
shRNA
Target Sets Individual Clones Gene Family Sets
Formats Available
  Bacterial Stock  
  Purified DNA Controls
  Lentiviral Transduction Particles
MISSION shRNA Product Overview
The library is currently provided as lentiviral transduction particles, purified plasmid DNA and frozen bacterial glycerol stock format harboring sequence-verified shRNA. Each gene is represented by a target set that consists of an average of 3-5 individual constructs or clones targeting different regions of the gene sequence. Bacterial cultures may be amplified from the glycerol stocks for use in purification of the shRNA plasmid DNA. Alternatively, the purified plasmid format does not require these extra steps as the growth and DNA purification are completed for you. Target cell lines may be transfected with the purified plasmid for transient or stable gene silencing (puromycin selection).
In addition, self-inactivating replication incompetent lentiviral particles can be produced in packaging cells (HEK 293T) by co-transfection with compatible packaging plasmids. Orders of 5 or less target sets (< 25 constructs) are provided in individual 1.4 ml TrakMates® tubes. Each TrakMates® tube contains a unique 2 dimensional barcode on the bottom of the tube that can be read using a corresponding reader. A printed value corresponding to The RNAi Consortium (TRC) clone number is also provided on each tube. Orders of >5 clone sets (>25 constructs) are automatically provided in a 96-well plate with a one dimensional barcode label on the plate and are provided with a CD containing plate map positions.
Manufacturing and Shipping Timelines
Each target set or individual clone is produced on demand in our state-of-the-art manufacturing facility. The following times are required to produce each format after the order appears in our LIMS manufacturing database:
SHGLY – Bacterial Stock 48 hours
SHDNA – Purified DNA 2 weeks
SHVRS – Lentiviral Transduction Particles 4 weeks
Orders ship Monday -- Thursday. Glycerol format orders received after 3 pm on Wednesday may not ship until the following Monday. Some orders may require additional time to process. Please allow up to 2 weeks additional shipping time for international orders.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
back to top
shRNA Homepage
RNAi Homepage