Fig. A. Actual Titer Measurements of Lentiviral Particles Manufactured by Sigma. This graph presents a representative sample of individual MISSION lentiviral particle preparations titered by our QC team. Viral particles are prepared in 96-well format using state-of-the-art robotics and are titered using the p24 assay. Sigma's minimum QC specification for a single preparation is 1 x 106 TU/ml.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
TrakMates is a registered trademark of Matrix Technologies Corporation.
back to top
back to Application Data
shRNA Homepage
RNAi Homepage
|