|
MISSION shRNA Lentiviral Transduction
Figure 1. Five individual clones from MISSION shRNA Target Set NM_002467 were co-transfected with a lentivirus packaging plasmid into HEK 293T cells. The resulting lentiviral particles were used to infect the human lung cancer cell line, H1299. 24 hours post-infection, 2.5 mg/ml of puromycin was added to select for infected cells. Infection efficiency was approximately 95%. 72 hours post-selection, cells were harvested. C-MYC message RNA level was detected by real-time PCR. Actin mRNA was used as an internal control.
Figure 2. Protein lysates from the shRNA infected cells were analyzed by western blotting using a c-MYC antibody. C-MYC protein levels corresponds to the mRNA knockdown shown in Figure 1.
Data generously provided by Dr. Steven B. McMahon, The Wistar Institute.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
TrakMates is a registered trademark of Matrix Technologies Corporation.
back to top
back to Application Data
shRNA Homepage
RNAi Homepage
|