|
Data courtesy of Sheila Stewart, Ph.D. Washington University School of Medicine
Figures A and B. Transduction efficiency and fluorescence intensity were compared between pRRL-eGFP lentiviral particles and MISSION™ TurboGFP™ lentiviral particles using increasing amounts of lentiviral supernatant. 5.0 x 105 normal BJ fibroblasts were transduced in 60 mm dishes. Fluorescence Activated Cell Sorting (FACS) was performed 60 hours post-transduction on 1.0 x 104 cells. Transduction efficiency of the MISSION TurboGFP Control Transduction Particles was higher compared to pRRL-eGFP particles. Fluorescence intensity was also much greater from the MISSION TurboGFP particles compared to pRRL-eGFP particles.
For more information on MISSION shRNA controls visit the MISSION shRNA Controls page.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
TrakMates is a registered trademark of Matrix Technologies Corporation.
back to top
back to Application Data
shRNA Homepage
RNAi Homepage
|