Functional Genomics and RNAi
Synthetic RNA
MISSION What's New
Key resources
RNAi Learning Center
shRNA
shRNA Search & Order
Learning Center
Promotions
Products
Labware
Academic Programs
Related Topics
siRNA
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Stability of pLKO.1-puro
shRNA
pLKO.1 Stability Assessed by Diagnostic Restriction Digestion
pLKO.1 Stability Assessed by Diagnostic Restriction Digestion
Digestion of pLKO.1 library constructs by PvuII produces three fragments of 3.8 kb, 2.5 kb, and 0.8 kb. PvuII digestions of 244 library clones in pLKO.1 were performed following ten rounds of sequential copying and regrowth. No evidence of recombination was observed. A representative digestion gel from a fraction of wells in a library plate from a 10th generation copy is shown.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
TrakMates is a registered trademark of Matrix Technologies Corporation.
back to top
back to Application Data
shRNA Homepage
RNAi Homepage