Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Synthetic RNA
MISSION What's New
Key resources
Pre-designed siRNA
RNAi Learning Center
shRNA
shRNA Search & Order
Library Information
Product Overview
Custom Services
Protocols, Data & FAQs
Metabolomics
Molecular Biology
Nutrition Research
Obesity Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Your Favorite Gene - Search
Life Science Quarterly
PathFinder
 LentiExpress™ Technology
shRNA
MISSION shRNA
LentiExpress™ RNAi Screens -
The Fastest Route to Viral-Based shRNA Gene Silencing.
Simply... Add Your Cells and Assay!
Would you like to conduct rapid, cost-effective lentiviral-based shRNA screens in primary or difficult to transfect cells? Are you interested in longer term or stable gene silencing while eliminating intensive reagent preparation of viral vectors and screen optimization parameters? Sigma Life Science provides an innovative solution with our LentiExpress technology.
LentiExpress screens consist of pre-arrayed lentiviral particles harboring shRNAs of the The RNAi Consortium collection. We offer this premier ready-to-use product format for the Human Kinase shRNA collection, so you can simply add your cells and proceed directly to loss-of-function screens!
  • Economical - Single use format
  • Simple, Streamlined Procedure - No viral particle preparation. Add your cells and go!
  • Save Time - Get your assay results 1 month earlier
  • Robust - Increased transduction efficiencies & no extra freeze-thaw cycles
  • Comprehensive - Average of 4-5 constructs per target gene
Go directly to:
Overview of The LentiExpress Method
LentiExpress Human Kinase shRNA Set
LentiExpress Optimization Plate
Data Using LentiExpress Technology
Ordering Information
Overview of The LentiExpress Method
LentiExpress Method image
LentiExpress vs. Standard Method image
back to top
New! MISSION® LentiExpress™ Human Kinase shRNA Set
Protein kinases are among the largest and most well studied gene families. Protein phosphorylation plays an essential role in intercellular communication in eukaryotic organisms by mediating signal transduction during development, transcription, immune response, metabolism, apoptosis, and cell differentiation. Aberrant regulation of kinases plays a causal role in many diseases, and the study of these proteins and their functions will contribute to the discovery and development of new therapeutics.
The MISSION LentiExpress Human Kinase shRNA Set contains 3109 lentiviruses carrying shRNA sequences targeting 673 human kinase genes allowing for quick, high throughput loss-of-function screens. Each gene is represented by a target set that consists of an average of 4-5 individual constructs, targeting different regions of the gene sequence. The particles are pre-diluted to approximately 5,000 viral particles per well in a single reaction volume of 30 microliters. In addition to the virus generated from the Human Kinases (K), each plate includes negative controls to monitor transduction efficiency.
  • Targets 673 human kinase genes
  • 3,109 lentiviral shRNA clones
  • Pre-arrayed in 41 ready-to-use 96-well plates
  • Up to 80 shRNA constructs per plate
  • Negative controls in outer columns
Representative Plate Layout image
back to top
New! MISSION LentiExpress™ Optimization Plate
Optimization of transduction conditions is a central component of successful delivery using the LentiExpress method. For maximal transduction and knockdown, it is important to determine the optimal ratio of virus to cells, or the MOI (multiplicity of infection), for each new cell type.
The LentiExpress Optimization Plate accelerates this process allowing rapid determination of the optimal cell density for transduction. The 96-well plate contains pre-arrayed aliquots of TurboGFP™ Control Transduction Particles, allowing the user to simply add the desired numbers of their cell type of interest. Efficiency of transduction is subsequently monitored by assessment of GFP fluorescence.
  • TurboGFP™ Control Particles for quick visual monitoring
  • Non-target shRNA Control Particles
  • Empty Vector Control Particles
Optimization Plate Layout image
back to top
LentiExpress Method for Screening Tumor Suppressor Gene Set.
The MISSION TRC Human Tumor Suppressor Gene Family Set (Catalog No. SH0531) was used as a proof of concept for the LentiExpress format. It was used to screen for genes that could enhance cell sensitivity to the widely used cancer therapy drug, Paclitaxel. Pre-arrayed lentiviral particles were used to transduce A549 cells in 96-well plates and knockdown the corresponding tumor suppressor gene expression. Cells were split into two samples. One sample set was treated with Paclitaxel and the second set received medium only. Results were normalized to pLKO.1-puro empty vector control virus (Catalog No. SHC001V). Outliers were observed that either conferred more resistance or sensitivity to Paclitaxel treatment.
Cell Growth Chart image
Gene Knockdown Chart image
Multiple clones for four genes confirmed the LentiExpress method. The clones conferred increased cell sensitivity to drug treatment. These genes have the potential to be prognostic markers in chemotherapeutic responses.
back to top
Ordering Information
Product #  Product Name
SHX001-1EA MISSION LentiExpress Human Kinases
SHX001-3EA MISSION LentiExpress Human Kinases (For triplicate experiments)
SHXC01-1EA MISSION LentiExpress Optimization Plate
SHXC01-3EA MISSION LentiExpress Optimization Plate
LentiExpress product image
RNAi Partnership Members may place orders by logging into our website with your institution's account number. Discounted pricing will be applied.
The LentiExpress products are in stock and ready for shipment 24 hours after your order is placed. Please note that these products are shipped on dry-ice and require immediate storage at -70°C.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a trademark of Sigma-Aldrich. Label License.
LentiExpress is a trademark of Sigma-Aldrich.
TurboGFP is a trademark of Evrogen.
back to top
shRNA Homepage
Functional Genomics & RNAi Homepage