|
When conducting experiments using MISSION TRC shRNA constructs, proper controls are a key element of experimental design to permit accurate interpretation of knockdown results and provide assurance of the specificity of the response observed. Sigma® offers a variety of positive and negative controls to monitor for efficient transduction, knockdown, and off-target effects.
All controls are available in both purified plasmid DNA and lentiviral particle format. The DNA format controls may be used in direct transfection-based experiments or may be used in conjunction with the MISSION Lentiviral Packaging Mix (SHP001) to create replication incompetent viral particles for use in transduction-based experiments.
|
Purified DNA |
| Product # |
Product Name |
Add to Cart |
| SHC001 |
MISSION® pLKO.1-puro Control Vector Purified DNA |
|
| SHC002 |
MISSION® Non-Target shRNA Control Vector Purified DNA |
|
| SHC003 |
MISSION® TurboGFP Control Vector Purified DNA |
|
| SHC004 |
MISSION® TurboGFP shRNA Control Vector Purified DNA |
|
| SHC005 |
MISSION® eGFP shRNA Control Vector |
|
| SHC007 |
MISSION® Luciferase shRNA Control Vector Purified DNA |
|
| SHC008 |
MISSION® shRNA Human Positive Control Vector #1 Purified DNA |
|
| SHC009 |
MISSION® shRNA Human Positive Control Vector #2 Purified DNA |
|
| |
|
Lentiviral Particles |
| Product # |
Product Name |
Add to Cart |
| SHC001V |
MISSION® pLKO.1-puro Control Transduction Particles |
|
| SHC002V |
MISSION® Non-Target shRNA Control Transduction Particles |
|
| SHC003V |
MISSION® TurboGFP Control Transduction Particles |
|
| SHC004V |
MISSION® TurboGFP shRNA Control Transduction Particles |
|
| SHC005V |
MISSION® eGFP shRNA Control Transduction Particles |
|
| SHC007V |
MISSION® Luciferase shRNA Control Transduction Particles |
|
| SHC008V |
MISSION® shRNA Human Positive Control Vector #1 Transduction Particles |
|
| SHC009V |
MISSION® shRNA Human Positive Control Vector #2 Transduction Particles |
|
Sigma's recommended controls for any shRNA experiment are provided in this Control Selection table and are closely aligned with the controls suggested in the
Nature Cell Biology editorial.1
| Recommended Control |
Objective |
Negative Control: Untreated Cells |
Untreated cells will provide a reference point for comparing all other samples. |
Negative Control: Empty construct, containing no shRNA insert
(SHC001V or SHC001)
|
MISSION pLKO.1-puro Control
The empty viral particles or DNA are a useful negative control that will not activate the RNAi pathway because it does not contain an shRNA insert. It will allow for observation of cellular effects of the transduction/transfection process. Cells transduced/transfected with the empty control provide a useful reference point for comparing specific knockdown.
|
Negative Control: Non-targeting shRNA
(SHC002V or SHC002)
|
MISSION Non-Target shRNA Control
This non-targeting shRNA is a useful negative control that will activate RISC and the RNAi pathway, but does not target any human or mouse genes. The short hairpin sequence cotnains 5 base pair mismatches to any known human or mouse gene. This allows for examination of the effects of shRNA transduction/transfection on gene expression. Cells transduced/transfected with the non-target shRNA will also provide useful reference for interpretation of knockdown. |
Positive Control: Positive reporter vector or lentiviral particles
(SHC003V or SHC003)
|
MISSION TurboGFP Control
This is a useful positive control for measuring transduction/transfection efficiency and optimizing shRNA delivery. The TurboGFP Control contains a gene encoding TurboGFP, driven by the CMV promoter. This control provides fast visual confirmation of successful transduction/transfection.
 |
Cells expressing TurboGFP 48 hours post-transduction. Lentiviral titer was estimated to be 1.0 x 106 TU/ml using a p24 based ELISA. |
|
Positive Control: Positive shRNA knockdown control
(SHC004V or SHC004) |
MISSION TurboGFP shRNA Control
This control contains shRNA sequence that targets TurboGFP expression. The shRNA has been experimentally shown to reduce TurboGFP expression by 99.6% in HEK293T cells 24 hours post-transduction. This control serves to quickly visualize knockdown in cells expressing TurboGFP. |
Positive Control: Positive shRNA knockdown control
(SHC005V or SHC005)
|
MISSION eGFP shRNA Control
This control contains shRNA sequence that targets eGFP expression (GenBank Accession # pEGFP U476561). The shRNA has been experimentally shown to reduce eGFP expression by 90% in C166-GFP mouse fibroblast cells 48 hours post-transduction by mRNA transcript level. This control serves to quickly visualize knockdown in cells expressing eGFP.
Knockdown efficiency of eGFP shRNA in HEK293T cells. Images shown 48 hours post-transfection. A) 100 ng of eGFP expression plasmid alone. B) 100 ng eGFP plasmid + 200 ng MISSION eGFP shRNA Control Vector. C) Bright field illumination.
|
| Back to top |
Reference
- Whither RNAi? Nature Cell Biology, 5, 489-490 (2003).
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
TurboGFP is a trademark of Evrogen Co.
back to top
shRNA Homepage
RNAi Homepage
|