Functional Genomics and RNAi
Synthetic RNA
MISSION What's New
Key resources
RNAi Learning Center
miRNA
shRNA
shRNA Search & Order
shRNA Learning Center
Promotions
Products
Labware
Academic Programs
Related Topics
siRNA
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 MISSION® shRNA Gene Family Sets
shRNA
The MISSION® TRC shRNA gene family sets are collections of genes related to specific functional classes. The format is designed for target discovery and pathway analysis. All of the gene family sets can be utilized without the need for robotics equipment. A dedicated researcher with a multichannel pipette can successfully utilize these gene family sets to discover novel small molecule targets and gene regulators. All gene family sets are pre-arrayed in 96-well plate formats, and are selected utilizing multiple databases including the NCBI, GO database, protein lounge, and Ingenuity. All human gene family sets are available as bacterial stocks, purified DNA, or lentiviral particles. The murine gene family sets are only available as bacterial stocks.
MISSION shRNA Gene Family Sets
Human
Gene Family
Sets
Product Numbers Product Information Supporting Products
Bacterial Stocks Purified DNA Lentiviral Particles Genes Clones* Antibodies Peptides and Purified Proteins siRNA Small Molecule Agonists and Antagonists
Apoptosis Pathway SH1911 SH1921 SH1931 437 2074 481 119 7396 420
B-Cell Activation SH2911 SH2921 SH2931 99 534 93 20 1419 126
Cell Adhesion Genes SH2211 SH2221 SH2231 366 1965 217 55 5942 100
Cytokine and Chemokine Receptors SH1311 SH1321 SH1331 97 459 69 17 1440 6
Cytokines and Chemokines SH0811 SH0821 SH0831 106 502 70 41 1405 50
Cytoskeleton Genes SH2311 SH2321 SH2331 273 1462 215 20 4588 65
DNA Repair Pathway SH1811 SH1821 SH1831 117 647 44 2 1815 44
Epigenetic Regulators SH3011 SH3021 SH3031 10 50 6 1 1593 2
Extracellular Matrix Genes SH2511 SH2521 SH2531 330 1657 3 1 200 58
G-Protein Coupled Receptors SH0211 SH0221 SH0231 479 1997 171 42 4842 52
Helicases SH2611 SH2621 SH2631 133 669 175 88 6002 1673
Ion Channels SH1011 SH1021 SH1031 285 1056 16 1 1700 0
JAK-STAT pathway SH2711 SH2721 SH2731 190 1123 152 101 4402 840
Kinases SH0111 SH0121 SH0131 622 2855 177 45 2697 104
Nuclear Hormone Receptors SH1111 SH1121 SH1131 46 209 292 67 9075 321
p53 Pathway SH2411 SH2421 SH2431 239 1410 24 2 880 218
Phosphatases SH0411 SH0421 SH0431 180 735 230 46 3952 139
T-Cell Activation SH2811 SH2821 SH2831 241 1262 14 0 2455 0
Tumor Suppressors SH0511 SH0521 SH0531 74 337 223 56 3597 142
Ubiquitin Hydrolases (DUBS) SH0711 SH0721 SH0731 112 615 60 4 1263 8
Ubiquitin Ligases (E1,E2,E3) SH2111 SH2121 SH2131 203 1018 72 11 1280 0
*The MISSION production and bio-informatics team constantly reviews and quality controls clones available for a gene family set. These numbers are very close to the actual number that will be shipped, but each researcher will receive a final plate map indicating the location and exact TRCN clone numbers.
back to top
Murine
Gene Family
Sets
Product Numbers Product Information Supporting Products
Bacterial Stocks Purified DNA Lentiviral Particles Genes Clones* Antibodies Peptides and Purified Proteins siRNA Small Molecule Agonists and Antagonists
Tyrosine Kinase SM2011 NA NA 109 518 17 6 3324 0
Phosphatase SM0411 NA NA 220 1110 109 28 1490 109
*The MISSION production and bio-informatics team constantly reviews and quality controls clones available for a gene family set. These numbers are very close to the actual number that will be shipped, but each researcher will receive a final plate map indicating the location and exact TRCN clone numbers.
back to top
Additional resources for ion channels research.
Additional sets will be made available over the coming months. If your gene set of interest is not yet available, please inquire for advanced custom preparation at RNAi@sial.com. We can assemble custom gene sets at any time.
Send this page to a colleague.
Contact Us
For questions about the library, pricing and quotes or other concerns, please e-mail us at: RNAi@sial.com.
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. Label License.
back to top
shRNA Homepage
RNAi Homepage