|
MISSION® Positive Control siRNA, designed using the Rosetta siRNA Design Algorithm, serve as an ideal complement to any siRNA silencing experiment. All MISSION® Positive Control siRNA have been validated to work*, so you can spend more time silencing your gene of interest and less time on optimizing internal controls.
Benefits of MISSION® Positive Control siRNA:
- Simplified optimization of siRNA transfection efficiency
- Increased target specificity, due to siRNA seed region optimization rules
- Validated gene silencing
- High quality siRNA produced at ISO 9000:2001 certified sites
- Freedom to operate for research use
| Available MISSION® Positive Control siRNA |
| Human |
| Product |
Gene |
| GAPDH |
glyceraldehyde-3-phosphate dehydrogenase |
| MAPK1 |
mitogen-activated protein kinase 1 |
| KIFF 11 |
kinesin family member 11 |
| Lamin A/C |
lamin A/C |
| Cyclophilin B (PPIB) |
peptidylprolyl isomerase B (cyclophilin B) |
| TP53 |
tumor protein p53 (Li-Fraumeni syndrome) |
*Mission Positive Control siRNA reduce the level of gene target by >75%, 24 hours after transfection with 30nM siRNA (Hela or 9L/lacZ cells).
Back to siRNA