Serum-Free Systems for MDBK and MDCK Epithelial Cells
by Terrell Johnson
Sigma-Aldrich Corporation, St. Louis, MO, USA
Introduction
Vaccines
developed from viruses grown in mammalian cell cultures are of major importance
in both human and animal health. The economic and sociological losses
due to viral infections in human populations and the animal-production
industry are significant, and necessitate the use of large-scale vaccination
programs. Therefore, efforts are continually underway to improve cell
culture systems that enhance our ability to increase the effectiveness
of vaccine production, to make the process more economical, and improve
the quality of life.
The
kidney epithelial cell lines, Madin-Darby Bovine Kidney (MDBK) and Madin-Darby
Canine Kidney (MDCK), are important and well-established culture systems
used to grow attenuated viruses for vaccine production.1, 2
The use of serum in a manufacturing process lessens the validity of data
interpretation, increases downstream-processing complexity, is less cost-effective,
and raises regulatory and safety concerns primarily related to adventitious
agents. To address these issues, we have developed four new media formulations.
These new formulations allow the growth of these cell lines to higher
cell densities than those cultured under traditional serum-supplemented
conditions.
Materials and Methods
All
materials were supplied by Sigma-Aldrich Corporation (St. Louis, MO) unless
otherwise stated.
Cell Lines
MDBK
(NBL-1, CCL-22) and MDCK (NBL-2, CCL-34) cell stocks were obtained from
ATCC (American Type Culture Collection, Manassas, VA). Stock cultures
of cells were grown in Dulbecco's Modified Eagle's Medium/Ham's F-12 (DME/F-12;
1:1) with 10% (v/v) fetal bovine serum (FBS). Cells were subcultured twice
per week following standard trypsinization protocols.
Serum-free culture system
Cultures
using the serum-free medium system required slightly modified protocols.
The following protocol was developed for use in maintaining cells in 25
cm2 T-flasks. Cultures were briefly washed with 0.2 ml/cm2
Dulbecco's Phosphate Buffered Saline (DPBS) containing calcium and magnesium
(Product Code: D8662. Cold 0.5% trypsin/5 mM EDTA (Product Code: T3924)
at a volume of 0.2 ml/cm2 was then added to the flask and allowed
to cover the culture surface for 30 to 60 seconds at room temperature.
After removal of the trypsin-EDTA, cultures were allowed to remain at
room temperature until cells became rounded and easily dislodged from
the surface by gently rapping the flask. Care was exercised to avoid excessive
force, as the cells were sensitive to mechanical damage. Cells were resuspended
in a 10 µg/ml solution of soybean trypsin inhibitor (Product Code:
T6414), at a volume of 0.2 ml/cm2 of original surface area.
Cells were pelleted at 500 rpm for 5 minutes, the trypsin inhibitor solution
removed, and the cells were resuspended in at least 0.1 ml of DPBS/cm2
of original surface area. Cells were then diluted into pre-warmed medium
and plated in 25-cm2 flasks at a minimum of 1-2 x 104
cells/cm2 of surface area.
Medium development
Medium
development was conducted using a modified Plackett-Burman method for
testing various supplements.3 Experiments were conducted by
plating cells at 1 x 104 cells/cm2 in 24-well multiwell
plates (Corning Costar, Cambridge, MA). Growth was monitored initially
by visual inspection of stained cells and later by either dye absorption
or direct measurement.
Since
MDBK and MDCK cell lines are frequently cultured under dynamic culture
systems, such as roller bottles and microcarrier beads in suspension,
it was critical to develop protocols for the use of these media under
these conditions. We employed 25 cm2 tissue culture flasks
on a rocker apparatus to mimic the shear forces associated with fluid
motion in dynamic culture systems. This model system was used to examine
the effect of medium components and optimize culture inoculation and handling.
Roller bottle cultures were inoculated at a density of 25,000 cells/cm2
in a volume of 0.2 ml/cm2 pre-warmed (37º C) medium to
ensure optimal distribution of cells. To facilitate cell attachment, initial
rotational speed did not exceed 0.2-0.3 revolutions per minute. After
12 to 24 hours, cells were firmly attached to the surface and speed was
increased to 0.5 revolutions per minute. Medium changes were performed
every 3-4 days to maintain optimal cell growth. To evaluate the performance
of the medium system using microcarrier beads in suspension, we used 100-ml
spinner flasks containing 75 ml of medium and 4 mg/ml beads. Beads were
inoculated with 25,000 cells/mg of beads to ensure that a sufficient number
of cells populated all of the beads. Use of a sufficient cell inoculum
to populate all the beads was vital since cells were not observed to migrate
from one bead to another. MDBK cell attachment and growth was evaluated
on dextran-based beads (Cytodex-1 and -2, Amersham Pharmacia Biotech,
Piscataway, NJ) and polystyrene-based beads (Plastic-Plus and Pronectin®-F
treated, SoloHill Engineering Inc., Ann Arbor, MI).
Cell freezing and Recovery after cryopreservation
Successful
freezing and recovery of MDBK and MDCK cells grown in these serum-free
formulations required modifications to standard techniques. Actively dividing
cells were trypsinized according to the protocol previously described,
pelleted by centrifugation, and resuspended at a concentration of 106
cells/ml in serum-free cell freezing medium (Product Code: C6295). Cells
were frozen in liquid nitrogen as previously described. Cells were recovered
by rapid thawing and seeded at a density of 40,000 cells/cm2
or higher in pre-warmed MDBK "recovery" medium or MDCK culture medium
without removal of the freezing medium. The "recovery" medium is a modification
of MDBK-GM, which supports a slower rate of cell growth than MDBK-GM and
thereby fosters improved cell recovery after thawing.
Cells
were allowed to attach for 2-4 hours, then the medium was removed and
replaced with fresh "recovery" or culture medium. MDBK cultures were maintained
for 2 or 3 days in "recovery" medium before the addition of MDBK-GM. For
MDCK cells, an equal volume of medium was added after 48 hours and a complete
medium change was performed on day 4 or 5.
Results and Discussion
Several
serum-free medium formulations, including Medium K-1 for MDCK cells4,
were examined for their ability to support MDBK and MDCK cells in both
static and dynamic culture systems. None of the formulas examined provided
results comparable to serum-supplemented formulations. We began our development
of a serum-free medium by investigating the effects of different basal
medium formulations on cell growth. A variety of different basal media
and various mixtures of these media were examined for their ability to
support growth with low levels of serum supplementation. Based on this
work, a mixture of DME and Ham's F-12 was selected as the basis of future
medium development.
The
most striking observations under serum-free conditions were the relatively
slow cell growth rates and the rapid formation of cellular domes or other
morphological changes. We sought to identify components that supported
growth and normal morphology of cells upon direct transfer from serum-supplemented
medium to serum-free medium. This was done using a modified Plackett-Burman
system and observing cellular morphology. Based on these observations,
we were able to identify several factors required for cell growth. Not
surprisingly, these factors normally provided by serum, included hormones
and growth factors. With the goal of making this medium regulatory friendly,
animal-derived proteins were eliminated as much as possible. All hormones
and growth factors used were recombinant proteins expressed in non-vertebrate
systems.
Analysis
of spent medium identified several amino acids and energy-producing compounds
that were used in disproportionate amounts or depleted from the medium.
We also observed a coordinate increase in other amino acids, such as alanine.
It has been reported that limitations of essential nutrients can result
in nutrient deficiency-induced apoptosis.5 To avoid this, we
then supplemented the medium with the requisite amino acids and analysis
of spent medium of the new formulations showed a more consistent profile.
Most amino acids were present in proportions similar to that of fresh
medium. Our observations revealed the need for adaptation of traditional
cell culture protocols including inoculation density, procedures to allow
cell attachment, and media exchange frequency in coordination with the
new medium formulations, to ensure optimal performance. The new medium
formulations included a growth medium for MDBK cells (MDBK-GM) which contained
a low level of animal-derived proteins (<100 µg/ml), a protein-free
medium (MDCK-PFM) and a low-protein, serum-free medium for MDCK cells
(MDCK-SF).
MDBK static and dynamic culture systems
Cells
cultured in MDBK-GM under static conditions showed minor transient morphological
differences in cell shape and vacuole formation when first transferred
to serum-free medium compared to cells cultured in serum-supplemented
medium (data not shown). Cells grew steadily, reaching maximal density
within 5-6 days, and then appeared to lose their ability to divide as
evidenced by an inability to be subcultured. This possibly was the result
of the cells entering quiescence or apoptosis. We have observed that MDBK
cells can undergo at least four passages in MDBK-GM medium without changes
in doubling time or cell morphology when subcultured during the growth
phase (data not shown).
While
MDBK-GM supported the rapid growth of cells to significantly greater densities
than serum-supplemented cultures, it did not stabily maintain these
densities as non-dividing cultures (dotted line in Figure
1). We reformulated MDBK-GM to minimize growth-inducing components
and reduce metabolically-stimulatory components. This reformulation allowed
cultures to reach levels sufficient to sustain cell viability at high
cell densities while minimizing cell replication. We called this new protein-free
medium MDBK Maintenance Medium (MDBK-MM). MDBK-MM supported the growth
of MDBK cells at a significantly reduced rate. By combining MDBK-GM and
MDBK-MM into a system for culturing MDBK cells, we achieved rapid growth
with no lag time and no weaning. By using MDBK-GM for 2-5 days and then
switching to MDBK-MM, an inoculum of 104 cells/cm2
in flasks or roller bottles expanded to 5 x 105 cells/cm2
in five days. This could be maintained for at least two weeks in both
static and dynamic cultures.
To
test the effectiveness of the MDBK medium system under dynamic culture
conditions, we evaluated growth performance in roller bottles and on microcarrier
beads in suspension. MDBK cultures were successfully established and maintained
in roller bottles. Within five days, cells inoculated at a density of
25,000 cells/cm2 achieved cell densities in excess of 5 x 105
cells/cm2. These densities were maintained for over 14 days
with medium changes every 3-4 days. We have also examined the growth of
cells on microcarrier beads. Using an inoculum of 25,000 cells/mg of beads,
we have been able to achieve uniform coverage of the beads with densities
of 5 x 105 cells/mg beads. The use of sufficient cell inoculum
to populate all the beads was vital since cells were not observed to migrate
from one bead to another.
MDCK static and dynamic culture systems
Cells
cultured in the serum-free static culture system grew steadily using either
serum-free, low-protein medium or protein-free medium and reached maximal
density within 4-5 days. We observed that MDCK cells could undergo at
least four passages in MDCK-PFM without changes in doubling time when
subcultured during the growth phase (data not shown). Cells exhibited
an inability to thrive in protein-free medium at low inoculation densities
and minimum inoculations of 10,000 cells/cm2 were required.
Cell
growth under dynamic culture conditions, which simulated roller-bottle
cultures, was also evaluated. We observed that while the protein-free
medium supported growth in static cultures, this medium did not provide
optimal support
to allow initial cell attachment under non-static conditions. During medium
development, it was observed that the addition of protein(s) to the medium
had a positive effect on the ability of MDCK cells to attach to the culture
surface under stressful conditions. A screening of proteins revealed that
several proteins could provide the necessary nutritional supplementation
to facilitate cell attachment at low cell densities in dynamic cultures.
From this, we determined that a final protein concentration of 100 µg/ml
was sufficient to allow cells to attach under dynamic culture conditions
while still expressing the desired growth characteristics. We refer to
this medium as MDCK low protein medium or MDCK- LPM.
MDCK-PFM or MDCK-LPM
media support the growth of cells with no lag time and no weaning in static
cultures. Additionally, roller bottle cultures can be initiated in MDCK-LPM
to facilitate cell attachment. The medium can be replaced after 4-5 days
with MDCK-PFM to minimize the amount of animal-derived protein present
in the final product ( Figure
2). Under these conditions, an inoculum of 2 x 104 cells/cm2
in flasks or roller bottles expands to 4 x 105 cells/cm2
in four days. These densities can be maintained for at least two weeks
in both static and dynamic cultures. We also found that cells grown on
microcarrier beads will achieve densities of 5 x 105 cells/mg
beads in 4-5 days with uniform coverage of the beads in cultures at an
inoculum of 25,000 cells/mg of beads.
Virus production
Both cell medium
systems were evaluated for their ability to support the production of
infectious viral particles. Both medium systems supported levels of viral
expression similar to or superior to cells grown in medium supplemented
with 10% FBS (Figures
3 and
4).
Conclusions
While the advantages
of reducing or eliminating serum and other animal-derived products in
medium used for manufacturing are obvious, the development of new medium
formulations are complicated by various limitations. In general, cell
lines have many nutritional requirements in common. However, each cell
line also possesses its own requirements. Many cell types, although similar,
may demonstrate different needs. The difficulty of medium development
is exemplified by contrast between MDBK and MDCK cell lines. While both
lines are derived from kidney epithelial cells, media developed for one
line does not adequately meet the requirements of the other. Similarly,
media developed for the culture of cells in static cultures may not give
satisfactory results when used in dynamic culture systems, such as roller
bottles and stirred cultures. Protocols employed for serum-supplemented
cultures did not give positive results when employed in serum-free systems.
Thus, the development of new culture media is not merely the modification
of existing formulas but rather the coordinate development of cell culture
systems of new or modified handling protocols that work in conjunction
with modified medium formulations.
We have developed
a serum-free medium system for MDBK cells (Product Codes: M3553 and M0682) that supports rapid cell growth and maintains high cell densities
for over 14 days through feeding of the cultures. For MDCK cells, we have
developed a low-protein, serum-free medium (Product Code: M3803) and
a protein-free medium (Product Code: M3678). Both cell lines have achieved
cell densities in excess of 5 x 105 cells/cm2, which
is comparable to medium supplemented with 10% FBS. Based on initial studies,
the viral productivity of cells grown under serum-free conditions appears
to be comparable to serum-supplemented cultures. Additionally, the ability
to maintain high cell densities for extended periods of time provides
the opportunity to combine this medium system with a "multi-harvest" process
such as that described by Noe et al.6 which further enhances
yield and reduces costs. These products offer a number of advantages over
traditional methods using serum-supplemented systems, such as improved
medium definition, reduced cost, improved downstream processing and fewer
regulatory concerns. The performance of our new media formulations combined
with the advantages previously cited offer the potential to significantly
improve the efficiency of batch manufacturing processes used for growing
viruses for use in vaccine production.
References
1. McRoberts, J.A.,
et al., The Madin-Darby canine kidney (MDCK) cell line, in Functionally
Differentiated Cell Lines (G. Sata, Ed.) pp. 117-139 (1981).
2. Gaush, C.R.,
et al., Characterization of an established line of canine kidney cells
(MDCK). Proc. Soc. Exp. Biolo. Med. 122, 931-935 (1966).
3. Plackett,
R.L., and Burman, J.P. The design of optimum multifactorial experiments.
Biometrika 34,255-272 (1946).
4. Taub, M.,
L., et al., Growth of Madin-Darby canine kidney epithelial cell (MDCK)
line in hormone-supplemented, serum-free medium. Proc. Natl. Acad. Sci.
USA 76, 3338-3342 (1979).
5. Al-Rubeai,
M. and Singh, R. P., Apoptosis in cell culture. Curr. Opin. Biotechnol.
9, 152-156 (1998).
6. Noe, W.,
et al., Optimization of vaccine production for animal health. Cytotechnology
15, 169-176 (1994).
About the Authors
Terrell Johnson, Ph.D., is a senior scientist in Cell Culture R&D at Sigma- Aldrich, St. Louis, MO.
| ORDERING INFORMATION |
| Product Code
|
Product Name |
Unit |
|
M3553
|
MDBK-GM Serum-free Medium
|
1 L
|
|
M0682
|
MDBK-MM Protein-free Medium
|
1 L
|
|
M3803
|
MDCK Serum-free Medium
|
1 L
|
| M3678 |
MDCK Protein-free Medium |
5 ml |
| G7513 |
L-Glutamine Solution (200 mM)
|
5 ml |
| |
|
10 ml |
| RELATED PRODUCTS |
| Product Code
|
Product Name |
Unit |
|
A1345
|
Anit-oxidant Supplement
|
5 ml
|
|
P8483
|
Polyamine Supplement
|
5 ml
|
| SUPPORTING LITERATURE |
MDBK/MDCK brochure
(CRT)
Media for Biotechnology/Products for Formulation brochure (DOJ)
Life Science Catalog 2000-2001, pages 353-354