One-Tube Preparation and PCR Amplification of DNA From Plant Leaf Tissue with Extract-N-AmpTM
by Carol Kreader, Scott Weber, and Keming Song
Sigma-Aldrich Corporation, St. Louis, MO, USA
Introduction
The
use of PCR amplification to detect target DNA sequences has many applications
in plant genotyping, gene mapping, diagnostics, and diversity assessment.
PCR itself is simple to set up, and requires little hands-on time. However,
most methods for preparing DNA from plant tissues are time consuming,
tedious, and labor intensive. Virtually all require mechanical disruption,
such as grinding in liquid nitrogen1 or reciprocal shaking2,
to break the plant
cell wall. Furthermore, many use multiple extraction steps with organic
solvents (phenol, chloroform), detergents (CTAB, SDS)1, salts
(NaCl, ammonium acetate),3 and/or polyvinylpyrrollidone (PVP)3
to remove polysaccharides and polyphenolic components that can inhibit
enzymatic reactions. Clearly, none of these methods is amenable to rapid
analysis with more than a few test samples.
We
have developed Extract-N-Amp, a one-tube extraction and PCR amplification
system, for DNA from plant leaves. Briefly, a 0.5 to 0.7 cm disk of leaf
tissue, cut with a standard paper punch, is incubated in Extraction Solution
at 95 °C for 10 minutes (Figure
1). After an equal volume of Dilution Solution is added to the extract
to neutralize inhibitory substances, the extract is ready for PCR. An
aliquot of the diluted extract is then combined with Extract-N-Amp PCR
Mix and user-provided PCR primers to amplify target DNA. The Extract-N-Amp
PCR Mixes are optimized specifically for use with the extraction reagents,
and contain JumpStartTM antibody for Hot Start PCR to enhance amplification
specificity. The REDExtract-N-AmpTM PCR Mix (XNA-P) contains REDTaqTM
to allow direct loading of the amplification product on an agarose gel
without separate addition of loading buffers and dyes. The Extract-N-AmpTM
PCR Mix in the XNA-P2 and XNA-R kits lacks the red dye for use with non-gel
detection methods in which the red dye interferes.
Materials
and Methods
All
materials were obtained from Sigma-Aldrich Corporation (St. Louis, MO)
unless otherwise noted. Leaf tissue and PCR primer sequences were graciously
provided by Monsanto (St. Louis, MO). PCR primers were obtained from Sigma-Genosys
(The Woodlands, TX).
DNA extraction
Leaves
were frozen at -70 :C, transported on dry ice, and stored at -70
:C. For use, leaves were thawed slightly, and kept on ice. Duplicate
0.5-0.7 cm disks were cut from each leaf with a standard one-hole paper
punch and transferred with forceps to a 2-ml microcentrifuge tube containing
100 5l of Extraction Solution. Paper punch and forceps were rinsed
with 70% ethanol between samples. Tubes were vortexed briefly to cover
the leaf disk with Extraction Solution, then incubated at 95 :C in
a dry heating block for 10 minutes. After the incubation, 100 5l
of Dilution Solution was added and the mixture vortexed briefly. Further
dilutions of the extract (1:5 and 1:10) were made with a 50:50 mixture
of the Extraction and Dilution Solutions. Four microliters of diluted
extract was added to PCR, and the remainder stored at 4 :C (without
removing the leaf disk).
PCR amplification
Each
PCR contained 10 5l of REDExtract-N-Amp PCR mix, 4 both
forward and reverse primers at 0.4 5M, and 4 5l of diluted leaf
extract in a final volume of 20 5l. Reactions were assembled at room
temperature (22-25 :C). PCR conditions were: 94 :C for 3 minutes,
then 35 cycles of 94 :C for 30 seconds, optimum annealing temperature
(45, 48, 53, or 58 :C) for 30 seconds, and 72 :C for 1.5 minutes.
The final cycle was followed by incubation at 72 :C for 10 minutes.
Eight microliters of each PCR product were loaded directly onto a 1.5%
agarose gel for electrophoresis.
Results and Discussion
The
Extract-N-Amp kits were tested using leaves from 5 crop species: corn,
cotton, soybean, potato (Figure
2, Figure
3, and Figure
4), and wheat (no data shown). In preliminary experiments, we demonstrated
that extracts from corn and wheat leaves could be added directly to PCR,
but extracts from potato leaves inhibited PCR unless they were diluted
at least 5-fold. Adding an equal volume of the proprietary Dilution Solution
to the extracted material relieved PCR inhibition without such extensive
dilution (data not shown). Therefore, addition of Dilution Solution to
the extract before PCR is part of the standard procedure (Figure
1; lanes labeled "std" on Figures 2,
3, and
4). Note that more than enough DNA is released from a 0.5-0.7 cm leaf
punch for PCR detection with the standard procedure. In fact, PCR from
a 1:10 dilution of the extract usually yields enough PCR product for detection
on agarose gels (Figures
3 and
4, lanes labeled "1:10").
Extract-N-Amp
performs well with both immature and mature leaves. In
Figure 2, DNA was extracted from leaves of corn plants at the 4- or
8-leaf stage, and from fully mature corn plants. All gave ample PCR product
with the standard procedure (std), and low, but detectable, amounts of
PCR product after a 10-fold dilution of the leaf extracts (1:10).
In
some situations, it may be inconvenient to prepare leaf samples by cutting
disks with a non-disposable punch that must be cleaned between samples.
As an alternative, pieces of tissue similar in size to the punches (0.5-0.7
cm in diameter) were torn from corn and cotton leaves (Figure
3, "tear"). For comparison, pieces of the same leaf were cut with
the hole punch ("punch") or crushed by grinding in liquid nitrogen ("grind").
All were extracted and target DNA sequence was detected by PCR according
to the standard procedure, or after further 5-fold or 10-fold dilutions.
Both the torn and punched leaf tissue gave plenty of target DNA, detected
even after 1:10 dilutions. In contrast, ground cotton leaf extracts gave
no PCR product unless they were diluted, indicating that these extracts
inhibit PCR. Concurrent tests with soybean leaves gave results similar
to corn (data not shown). Therefore, either torn or punched pieces of
leaf tissue, 0.5-0.7 cm in diameter, may be used with Extract-N-Amp.
The
DNA extracted from leaves with Extract-N-Amp is quite stable in the Extraction/Dilution
Solution mixture at 4 :C. For
Figure 4, leaf extracts were stored at 2-8 :C for nine (cotton
and soybean) or ten months (potato), and then analyzed by PCR. All contained
DNA that was readily detected by PCR, even after 1:10 dilutions.
In
addition to the 5 crop species used at Sigma to test the kit, other researchers
have shown that Extract-N-Amp performs well with a variety of plant species.
Figure 5 shows successful extraction and PCR amplification with leaves
from Virginia creeper, Honeysuckle, Cottoneaster, Ficus benjamina,
and Cannabis sativa.
Conclusions
Extract-N-Amp performs well with mature as well as immature plant leaves. Torn leaf
tissue may be used instead of leaf punches, as long as the size is kept
at ~ 0.5-0.7 cm. The leaf extracts are stable for at least 9 months at
2-8 :C. Furthermore, leaves from a wide variety of plant species
may be used with Extract-N-Amp.
Acknowledgements
We
thank Kim Beazley and Rachel Krieb (Monsanto, St. Louis, MO, USA) for
providing leaf tissue and PCR primer sequences and John Backus (Sigma
Chemical Co R&D, St. Louis, MO, USA) for optimizing annealing temperatures
for the PCR primers. We also thank Andy Hopwood (Forensic Science Service,
Birmingham England) for allowing us to print his unpublished results ( Figure
5).
References and Note
1.
Ausubel, F., Brent, R., Kingston, R.E., Moore, D.D., Seidman, J.G., Smith,
J.A., and Struhl, K., Short Protocols in Molecular Biology, pp. 2.9-2-11
(John Wiley & Sons, Inc., New York, NY, 1997).
2. Geuna, J., Hartings, H., and Scienza, A., Plant DNA extraction based
on grinding by reciprocal shaking of dried tissue. Anal. Biochem., 278,
228-230 (2000).
3. Kim, C.S., Lee, C.H., Shin, J.S., Chung, Y.S., and Hyung, N.I., A simple
and rapid method for isolation of high quality genomic DNA from fruit
trees and conifers using PVP. Nucleic Acids Res., 25, 1085-1086
(1997).
4. The Extract-N-Amp PCR Mix is specifically formulated for use with the
Extract-N-Amp Extraction Solution. Use of a PCR mixture other than the
Extract-N-Amp PCR Mix will likely result in failure of the PCR.
About the Authors
Carol
Kreader, Ph.D., is a principal scientist and Scott Weber, B.S., is a research
associate in Nucleic Acid Purification R&D at Sigma-Aldrich, St. Louis,
MO. Keming Song, Ph.D., is a research fellow in Recombinant Protein Expression
R&D at Sigma-Aldrich, St. Louis, MO.
| ORDERING INFORMATION |
| Product Code
|
Product Name |
Unit |
| XNAP
|
REDExtract-N-AmpTM
Plant PCR Kit
(100 Extractions & Amplifications)
|
1 Kit
|
| XNAP2 |
XNAP2 Extract-N-AmpTM Plant PCR Kit
(100 Extractions & Amplifications) |
1 Kit
|
| XNAR |
XNAR Extract-N-AmpTM Reagent Kit
(1,000 Extractions & Amplifications)
|
1 Kit