The GenEluteTM Endotoxin-free DNA Purification
Kit: Pure DNA for Endotoxin-Sensitive Applications
by Fuqiang Chen, Scott Weber and
Jo Anne Kerschner
Sigma-Aldrich Corporation, St. Louis,
MO, USA
Introduction
Endotoxins are lipopolysaccharides
(LPS), found among outer cell-membrane components of Gram negative bacteria
such as E. coli. One E. coli cell contains approximately
2,000,000 LPS molecules. In mammalian systems, endotoxins are pyrogenic,
causing fever and endotoxic shock syndrome. The potential for contamination
of purified plasmid preps, raised in E. coli, by endotoxins is a
major concern in gene therapy. Additionally, endotoxins can significantly
reduce transfection efficiencies in endotoxin-sensitive cell lines such
as COS-7, HEK293, and CHO-K1.1 Other applications affected to
a lesser extent are restriction endonuclease digestion, cloning, sequencing,
and PCR.
LPS molecules are negatively charged
like DNA and are often co-purified with plasmid DNA. Both molecules behave
similarly on a silica surface or in anion exchange chromatography. The
density of endotoxins in CsCl ultracentrifugation is similar to that of
the plasmid-ethidium bromide complex, causing contamination of CsCl-purified
DNA. Purified plasmid DNA prepared by silica-based plasmid isolation systems
may contain extremely high levels (>1000 EU/µg plasmid DNA) of endotoxins.1
Plasmid DNA prepared by anion exchange chromatography or two rounds of
CsCl ultracentrifugation contains lower, but still significant levels (>4
EU/µg DNA) of endotoxin.2 In order to produce material
that is endotoxin-free, additional reagents and steps are required. This
can be accomplished either during or after DNA extraction.
Materials and Methods
All materials were supplied by Sigma
Chemical (St. Louis, MO) unless otherwise stated.
The GenEluteTM Endotoxin-free Plasmid
Midiprep and Maxiprep kits provide a simple, rapid, and cost-effective
method for isolating endotoxin-free plasmid DNA from recombinant bacterial
cultures. An endotoxin removal step is integrated into the GenEluteTM plasmid
purification system. After endotoxin removal, plasmid DNA is purified on
a silica-based membrane in an easy-to-use spin-column format. Up to 120
µg of plasmid DNA free of endotoxin contamination (<0.1 EU/µg
DNA determined by Limulus Amebocyte Lysate kinetic test) can be recovered
from 5-40 ml of overnight bacterial culture with the Midiprep kit. Up to
1.2 mg of plasmid DNA free of endotoxin contamination (<0.1 EU/µg
DNA) can be recovered from 25-130 ml of overnight bacterial culture with
the Maxiprep kit.
Recombinant E. coli cells are
harvested from an overnight culture by centrifugation and subjected to
a modified alkaline-SDS lysis procedure to produce a cleared lysate. Chromosomal
DNA and other cellular components are precipitated and removed from the
lysate by centrifugation. Endotoxins are removed from the cleared lysate
by extraction with a temperature-mediated two-phase separation. The blue
dye in the lower organic phase allows easy location of the interface and
facilitates collection of the clear upper phase containing the plasmid
DNA. After the addition of a DNA-binding solution, the endotoxin-free cleared
lysate is loaded onto a midi- or maxi-binding column that is seated inside
a collection tube. Plasmid DNA is selectively bound onto the silica membrane
when the lysate flows through the column during centrifugation. Salts and
other contaminants are removed from the column by a spin-wash step. DNA
is eluted in endotoxin-free water.
Results and Discussion
Plasmid DNA purified by this method
is predominately in its super-coiled form. The 260/280 nm ratio is consistently
between 1.8 and 2.0. There is no evidence of genomic DNA or RNA contamination
(determined by agarose gel electrophoresis). The level of residual endotoxins
in purified DNA is consistently <0.1 EU/µg plasmid DNA determined
by Limulus Amebocyte Lysate kinetic test. Purified plasmid DNA is ready
for use in cell transfection and other downstream applications, such as
restriction endonuclease digestion, ligation, sequencing or PCR.
It takes approximately 95 minutes to
purify endotoxin-free plasmid DNA using the Midiprep kit and
105 minutes for the Maxiprep kit.
This compares favorably with other commercially available kits (Figure 1). Endotoxin-free plasmid DNA purified using the GenEluteTM Endotoxin-free
kits cuts efficiently with restriction enzymes. Figure
2 shows the action of Hind III and EcoRV restriction
enzymes on the plasmid DNA (pCMV-SPORT-ßgal) when 1 µg was
incubated with the enzymes at 37 ºC for 2 hours. Each lane was loaded
with half of a digestion.
Figure 3 demonstrates the transfection efficiency of plasmid DNA prepared
with Sigma's GenEluteTM Endotoxin-free kits versus those of competitors.
Plasmid DNA was prepared according to manufacturers' instructions. The
data show the average of six replicates for each plasmid isolation method.
All transfections were in CHO-K1 cells. The OD measurements were taken
at 420 nm and the units of ß-galactosidase per plate were determined
using the ß-Galactosidase Reporter Gene
Activity Detection kit (ß-Gal; Product Code: GAL-A). The cells were
grown to 60-75% confluency. Cells were transfected using 3 µg of
plasmid DNA per 15 µl of Escort IV, a transfection reagent (Product
Code: L 3287). The ß-Gal kit was used after a 68-70 hour post-transfection
incubation.
Conclusions
The GenEluteTM Endotoxin-free Plasmid
Midiprep and Maxiprep kits compare favorably with other commercially-available
kits. Yield for the Midiprep kit is up to 120 µg and up to 1.2 mg
for the Maxiprep kit. The transfection efficiency is superior compared
to competitors. Purified DNA is compatible for other downstream applications such as restriction endonuclease digestion,
sequencing, and PCR.
References
1. Weber, M., et al., Effects of lipopolysaccharide
on transfection efficiency in eukaryotic cells. BioTechniques, 19,
930-40 (1995).
2. Cotten, M. et al., Lipopolysaccharide
is a frequent contaminant of plasmid DNA preparations and can be toxic
to primary human cells in the presence of adenovirus. Gene Therapy, 1,
239-246 (1994).
3. Birnboim, H.C., A rapid alkaline
extraction method for the isolation of plasmid DNA. Methods Enzymol., 100,
243-55 (1983).
4. Boom, R., et al., Rapid and simple
method for purification of nucleic acids. J. Clin. Microbiol., 28,
495-503 (1990).
5. Ehlert, F., et al., Importance
of DNA quality for transfection efficiency. BioTechniques, 14, 546
(1993).
GenElute is a trademark of the Sigma-Aldrich
Corporation.
About the Authors
Fuqiang Chen, Ph.D., is a senior scientist
and Scott Weber, B.S., is an associate scientist in NAP, PCR and Sequencing
R&D at Sigma-Aldrich, St. Louis, MO. Jo Anne Kerschner, Ph.D., is the
manager of NAP, PCR and Sequencing R&D at Sigma-Aldrich, St. Louis,
MO.
| ORDERING
INFORMATION |
| Product Code |
Product Name |
Unit |
| PLED35 |
GenEluteTM
Endotoxin-free Plasmid Midiprep kit |
1
kit (35 reactions) |
| PLEX15 |
GenEluteTM
Endotoxin-free Plasmid Maxiprep kit |
1
kit (15 reactions) |
| SUPPORTING
LITERATURE |
| For
additional information, request technical bulletins (PLED 35) and
(PLEX 15) Nucleic Acid Brochure (DHB) |
|