Applications with REDTaq
by Brian Ward and Keming Song
Sigma-Aldrich Corporation, St. Louis, MO, USA
Introduction
Since its introduction in the Fall of 1998, REDTaq has been well received
by the scientific community. The original product idea was to add convenience
to PCR by formulating a reaction that also contained loading buffer and
dye. The end product would allow the researcher to see which samples contained
enzyme and would remove one step in post-PCR product analysis, but would
not hinder any conceivable post-PCR uses for the amplified DNA. The product
name, REDTaq, is meant to reflect simplicity of use to the customer; the
enzyme would work like Taq polymerase in every other way, but have
color.
Results and Discussion
REDTaq in downstream applications
From the myriad of chromophores that could have potentially fulfilled
the above objectives, one was selected to provide an amplification system
that performed as well as our standard Taq DNA polymerase. Included
in the screening were PCR and post-PCR concerns including PCR toxicity,
PCR product purification methods, restriction digests, ligation, and transformation.
One of the most stringent tests of the screening was PCR yield. As can
be seen in Figure 1, the PCR yields with the newly formulated product are comparable
to those of Taq. Additionally, PCR products from a REDTaq reaction
mixture can be used in automated sequencing without causing fluorescent
artifacts or hindering the sequencing reaction or length of read. An unforeseen
benefit of REDTaq is that one can easily see if a reaction has been thoroughly
mixed. All in all, REDTaq performs as we first envisioned. Aside from
the added conveniences of visualization and direct loading, the additives
have no effect on the reaction.
Compatibility of REDTaq in long and hot start PCR
Since the initial formulation of REDTaq, we have found the formulation
to be completely inert when stored as a PCR master mix. This has allowed
the recent release of REDTaq ReadyMixes. Slight optimizations of the initial
formulation have been shown compatible with Taq blends (AccuTaq)
and Taq directed antibodies (JumpStart). In Figure
2 the compatibility of REDTaq with hot start antibody is demonstrated
by showing the effect of REDTaq with and without antibody when template
amounts are low. Figure
3 shows that JumpStart REDAccuTaq is comparable
or better than the most robust blends offered by competitors.
Optimizing REDTaq PCR
As REDTaq is essentially identical in performance with Taq, REDTaq
and Taq PCR optimization are operationally identical. However,
REDTaq is different from Taq in its magnesium requirements. Although
methods of optimizing magnesium concentration for REDTaq and Taq
will be unchanged, one should be aware that magnesium concentrations optimized
for Taq may not be optimal for REDTaq, even if all other parameters
(template, primers cycling conditions etc.) are the same.
PCR optimization can usually be effected by changing one or more of
the following (in order of importance): template quality, primer design,
cycling parameters and magnesium concentrations. There are many excellent
reviews covering PCR optimization,1,2 and most good technical
libraries will include recent books covering general PCR techniques.3,4
Most troublesome PCR can be made to work by changing one or more of the
parameters described previously. However, this is far from a general method
for PCR optimization. If template quality is sufficient, then the problem
is often due to the complexities involved in PCR annealing, which is a
function of the melting temperature (Tm). Tm
is the temperature at which the annealing and dissociation rates of the
primer: template complex are equal. Dissociation is unimolecular and therefore
independent of concentration, while annealing is bimolecular and dependent
on PCR product and primer concentrations. To put it simply, Tm
values change during PCR. One technique, which allows a certain amount
of error in Tm calculation, is 'Touchdown' PCR.5
In this method, the annealing temperature is set five to ten degrees above
the calculated Tm. Annealing temperatures of subsequent
cycles are slowly dropped over the course of cycling. Touchdown PCR forces
the initial amplification steps to occur at the most stringent annealing
conditions possible, with subsequent, less stringent amplification relying
on the mass action of the correct amplicon to outcompete any side products.
This strategy is useful in that it is a ubiquitous means to get amplification.
It is worth trying after a positive control experiment has demonstrated
there are no PCR reagent problems.
Conclusion
REDTaq DNA polymerase with its optimized reaction buffer performs identically
and can directly replace Taq DNA polymerase in most applications.
The exception to this is with troublesome PCR. The difference becomes
a concern when using non-standard magnesium concentrations.
References
1. Linz, U., Protocol optimization and the need for standardization of
the polymerase chain reaction. Meth. Mol. Cell. Biol., 2, 98-102
(1991).
2. Roux, K. H., Optimizing and troubleshooting in PCR. PCR Methods Appl.,
4, S185-94 (1995).
3. Innis M. A and Gelfand, D. H, PCR protocols: a guide to methods
and applications, pp. 3-20, (Academic Press, New York, 1990).
4. Powell, S. J., PCR: essential data, pp. 72-86 (Wiley & Sons,
New York, 1995).
5. Don, R. H. et al., 'Touchdown' PCR to circumvent spurious priming during
gene amplification. Nucl. Acid Res., 19, 4008 (1991).
REDTaq , JumpStart, AccuTaq and JumpStart REDAccuTaq are trademarks of
the Sigma-Aldrich Corporation.
About the Authors
Brian Ward, Ph.D., is a principal scientist in PCR R&D and Keming
Song, Ph.D., is a research fellow in Recombinant Protein Expression R&D
at Sigma-Aldrich, St. Louis, MO.
| ORDERING
INFORMATION |
| Product Code |
Product Name |
Unit |
| D4309 |
REDTaq
DNA Polymerase |
50 units |
| |
|
250 units |
| |
|
2,500 (10 x
250) units |
|
D2812
|
REDTaq
DNA Polymerase without MgCl2 |
50 units |
| |
|
250 units
|
| |
|
2,500 (10 x 250) units
|
|
D8312
|
REDTaq
Genomic DNA Polymerase |
50 units |
| |
(Enhanced Amplification
on Genomic templates)
|
250 units |
| SUPPORTING
LITERATURE |
| PCR
Brochure (BKR) |
|