A New Medium for Adenovirus Production
By Terrell Johnson, Teena Rull, Brad Fuhr, Brigid DeLong, and Matthew
Caple.
Sigma-Aldrich Corporation, St. Louis, MO, USA
Introduction
The use of recombinant DNA technology to restore defective gene function
(gene therapy) offers the potential for improving the lives of individuals
afflicted with inborn errors of metabolism. Many of the vectors being
investigated for use in gene therapy are based on viruses, with adenoviruses
being one of the most commonly used delivery systems. Several helper cell
lines are employed for the production of gene therapy vectors derived
from adenoviruses. One of the inherent problems associated with many of
the cell line-vector combinations used for gene therapy is the generation
of wild-type replication competent adenoviruses (RCAs). The generation
of RCAs results from recombination between the vectors and viral sequences
in the helper cells used to grow the vector that can contaminate a vector
preparation intended for therapeutic use. One approach to eliminating
this problem is the construction of vector-helper cell combinations that
do not contain overlapping viral sequences eliminating the potential for
homologous recombination between the vector and viral sequences in the
helper cells. The Per.C6® helper cell line is engineered to contain
a specific sequence from the adenovirus serotype 5 (Ad5) E1A- and E1B-encoding
regions. The use of this cell line in combination with vectors which have
the E1 region deleted to remove overlapping sequences eliminates the concern
of generating RCAs by recombination. Similarly, the possible presence
of adventitious agents associated with animal-derived components frequently
used in cell culture medium also represents a potential source of contamination
of products produced using cell culture systems. These concerns have spurred
the development of animal component-free protein-free media for use in
manufacturing. With these objectives in mind, Sigma developed a medium
optimized for the growth of Per.C6® cells and designed
to meet the regulatory needs of manufacturers for the production of biotherapeutic
agents.
Materials and Methods
All materials were supplied by Sigma-Aldrich Corporation (St. Louis, MO)
unless otherwise stated.
Per.C6 ® cells are engineered to contain a specific portion of the
E1 region of adenovirus serotype 5 (Ad5) to enable this cell line to support
the replication of adenovirus-derived vectors deleted for the E1 region.
Per.C6® cells were cultured as attached cells in T-flasks in DMEM
+ 10% FBS (Product Code F2442). Agitated small-scale suspension cultures
were grown in 125 ml-1 L spinner flasks in protein-free medium on magnetic
stirrer platforms at 50-60 rpm. Cells were seeded into medium at 2.5 x
105 viable cells/ml and incubated at 37 °C in a humidified atmosphere
of 5% CO2. Total cell counts were determined using an electronic
particle counter. Cell viability was estimated by trypan blue dye exclusion.
Cells were grown in protein-free medium for multiple passages and did not
require adaptation prior to evaluation of growth and virus production in
batch culture. Cells grown in serum-supplemented medium were adapted to
protein-free medium using the following regimen. When cells cultured in
serum-supplemented medium reached approximately 80% confluence, the medium
was removed and replaced with protein-free medium and the flask incubated
overnight. The following day the flask was rapped sharply against the palm
of the hand to dislodge cells into suspension and the cells were collected
by centrifugation (200 x g for 5 minutes). Only cells that went into suspension
readily were collected. Cells were never released by mechanical scrapping
or enzymatic action. Cells were resuspended in fresh protein-free medium
at 5-8 x 105 viable cells/ml and transferred to an appropriately
sized T-flask. The cultures were counted daily and when the cell titer reached
1-1.2 x 106 cells/ml fresh medium was added to reduce the titer
to 5-8 x 105 /ml. When the number of cells was sufficient to
seed a minimum of 75 ml of medium at 3-4 x 105/ml the cultures
were transferred to a spinner flask at 50 rpm. Suspension cultures were
maintained by initiating new cultures at 2.5 x 105 viable cells/ml
when the cell titer reached 1-1.5 x 106 cells/ml.
Virus production
Viral production experiments were set up in singlet by inoculating Per.C6®
cells in 100-ml spinner flasks at a density of 2.5 x 105 cells/ml.
After three days of growth at 37 °C and 5% CO2, the spinners
were infected with 3 x 109 virus particles (moi 3)
of the rAd5 containing the ß-galactosidase (ß-gal) reporter
gene (Crucell, Leiden, The Netherlands). The infected cells grew for an
additional three days before collecting the virus for quantification. The
collection process consisted of centrifuging each infected culture sample
at 200 x g for five minutes. The supernatant was removed and the pellet
was resuspended in 10 ml of DPBS + 10% glycerol. Both supernatants and cell
lysates were stored at 70 °C. To quantify the virus concentrations,
a freeze/thaw process was performed three times to lyse the virus from the
cells. The freeze/thaw process consisted of freezing the samples at 70
°C and immediately thawing at 37 °C. The samples were centrifuged
at 200 x g for five minutes and the supernatant was drawn off for virus
titration. For virus titrations, 96-well plates were set up by inoculating
each well with 0.100 ml of attached Per.C6® cells (2.0 x 105
cells/ml) in DMEM + 4 mM glutamine + 10% FBS and incubated for three days
at 37 °C and 5% CO2. When setting up the virus titrations,
six ten-fold serial dilutions were prepared from each cell lysate (105
to 1010) in DMEM + 4 mM glutamine + 10% FBS. Each dilution
infected one column (eight wells) of a 96-well plate by adding 0.100 ml/well.
A control column of wells was set up with 0.100 ml of DMEM + 4 mM glutamine
+ 10% FBS. The infected plates were incubated for six days at 37 °C
and 5% CO2. This 96-well plate design was set up for high-throughput
analysis for use with ß-galactosidase assay.
ß-galactosidase assay
ß-gal activity was assayed using a colorimetric ß-galactosidase Reporter
Gene Activity Detection Kit (Product Code GAL-A). After incubation of 96-well
plates, the medium was removed from the infected wells and 100µl of
cell lysis buffer (1X) was added to each well. After a 20-minute incubation
at 37 °C, 100 µl of ß-gal assay buffer (2X) was added to each
well. The plates incubated for 30 minutes at 37 °C. Next, a plate reader
was used to determine absorbency values at a wavelength of 405 nm. The absorbency
readings were applied to the tissue culture infectious dose (TCID50)
equation to obtain quantitative data.
Quantitation of virus production
The tissue culture infectious dose is defined as the dilution of sample
at which 50% of the replicate cell culture inoculated with the sample becomes
infected. The method for infectious virus quantitation as described by Karber4
was used in calculating the TCID50 value: -m = log10
starting dilution [p-0.5] x d. The equation is defined where m is
the log10 TCID50 (per unit volume inoculated per replicate
culture), d is the log10 dilution factor, and p is the proportion
of wells positive for viral infection. The number of positive wells came
from the data obtained by the ß-gal assay. Positive wells are defined
as having optical densities greater than 0.500.
Results
Frozen cell stocks of Per.C6® cells were recovered by culture in serum-supplemented
medium and then adapted to growth in suspension culture in serum-free medium
by the methods described. The cell growth characteristics of cultures established
from multiple vials of frozen cell stocks and those adapted to suspension
culture were compared to assess variability introduced due to selection
during the recovery and adaptation processes. As seen in
Table 1, the cells adapted quickly to suspension culture without
significant changes in the population doubling time (PDT) observed with
attached cultures in serum-supplemented medium (approximately 26 hours)
and behaved consistently over multiple passages.
| Table 1. |
Population Doubling Time (Hours) |
| |
Passage |
| |
1 |
2 |
3 |
4 |
5 |
6 |
7 |
8 |
Average |
| Stock |
|
| 1 |
27.5 |
29.7 |
55.8 |
25.4 |
27.5 |
32.3 |
|
|
33.0 |
| 2 |
24.9 |
23.9 |
20.8 |
21.8 |
|
24.6 |
24.5 |
23.1 |
23.4 |
| 3 |
|
27.2 |
30.2 |
25.6 |
22.6 |
25.4 |
22.5 |
23.4 |
25.3 |
| 4 |
28.2 |
28.3 |
33.5 |
|
30.0 |
| 5 |
25.9 |
19.1 |
22.7 |
31.1 |
|
47.9 |
23.8 |
32.0 |
28.9 |
| 6 |
|
30.7 |
26.6 |
25.7 |
22.7 |
25.3 |
22.0 |
23.0 |
25.2 |
Initial studies indicated that cell populations grew to a maximum titer
of 2-3 x 106 cells/ml with high viability and then exhibited
very rapid decreases in viability. Studies were undertaken to understand
the effects of key components on cell growth and viability. Based on these
results, the original formulation was modified to produce a medium capable
of supporting high cell titers and allowing the cultures to maintain high
viability several days after reaching peak titer. Studies were then undertaken
to compare the performance of the new formulation Gene Therapy Medium (GTM-3;
Product Code G9916), with other commercially available media. The results
of cell growth studies of Per.C6® cells seeded at 2.5 x 106
cells/ml in 100 ml spinner cultures are shown in (Figure 1). These indicate that cells grown in our modified medium
(GTM-3) reached higher cell density ( > 4.0 x 106 viable cells/ml)
than cells grown in any of the other media tested. Similar results were
also observed in cultures grown in roller bottles (Figure 2)
To confirm that the modified medium formulation also supports adenovirus
production, a virus production was tested in the same group of media used
in the cell growth test. Virus production was tested as described in Materials
and Methods using a ß-gal colorimetric assay. Three days post infection,
GTM-3 resulted in a virus titer of 1.5 x 109 particles/ml and
Gene Therapy Medium 1 (GTM-1) resulted in a virus titer of 5 x 108
particles/ml, which is a three-fold increase in virus production (Figure 3). The same three-fold increase in virus production was also replicated
in roller bottles (Figure 4). GTM-3 supported a virus titer of 2.37 x 108
particles/ml compared to the GTM-1 virus titer of 7.50 x 107 particles/ml
(data not shown). These data suggest that Per.C6® cells produce more
virus when growing in the GTM-3 than in GTM-1 or any of the competitor media
(Table 2).
| Table 2. Comparison of virus production
from Per.C6® cells in GTM-1 and GTM-3. |
| |
|
|
| Experiment |
Medium |
Fold Difference |
Roller Bottle Assay |
GTM-3 |
3.2 |
| |
GTM-1 |
1 |
Spinner Flask Assay |
GTM-3 |
3 |
| |
GTM-1 |
1 |
| Spinner Flask Assay |
|
|
| 2 days post infection |
GTM-3 |
2.9 |
| |
GTM-1 |
1 |
3 days post infection |
GTM-3 |
2.8 |
| |
GTM-1 |
1 |
4 days post infection
|
GTM-3 |
3 |
| |
GTM-1 |
1 |
Conclusions
As biotechnology and pharmaceutical manufacturers strive to optimize downstream
processing and meet increasingly stringent regulatory guidelines, development
of new media to meet their needs must focus on two distinct and sometimes
divergent goals. First, animal-derived components must be eliminated due
to current and future regulatory concerns regarding raw materials used during
the manufacturing of injectable biotherapeutic agents. Second, without serum,
cell growth and productivity over a prolonged duration must be similar or
exceed that of serum-containing medium. Additionally, it is often desirable
to adapt adherent cell lines to suspension culture to facilitate the manufacturing
process. Meeting these goals, we have developed a medium for Per.C6®
cells, GTM-3, that is optimized for cell growth in suspension culture and
maximizes virus production. The medium meets the need of biotechnology and
pharmaceutical manufacturers for production of adenoviral vectors using
the paired system of rAd5 vector and Per.C6® cells and has the following
characteristics:
- Free of proteins and other animal-derived components
- Cells require little or no adaptation.
- Cultures consistently grow to a density of four to five million cells/ml
versus two to three million cells/ml in other commercially available media.
- The medium facilitates a metabolic shift in the cells to allow the
cells to maintain high viability for several days after reaching maximum
cell density.
- Consistent growth of cultures for greater than two weeks.
- The medium supports viral production three times greater than cells
grown in other media.
References
-
Fallaux, F.J., et al., New helper cells and matched early region 1-deleted
adenovirus vectors prevent generation of replication-competent adenoviruses.
Human Gene Therapy 9, 1909-1917. (1998).
-
Hehir, K.M., et al., Molecular characterization of replication-competent
variants of adenovirus vectors and genome modifications to prevent their
occurrence. J. Virol. 70, 8459-8467 (1996).
-
Karber, J., Beitrag zur kollektiven Behandlung pharmakologischer Reihenversuche-(On
collective treatment of serial pharmacologic studies). Arch. Exp. Pathol.
Pharmakol. 162, 480-483 (1931).
-
Monica, T.J., et al., Monitoring adenovirus infections with on-line and
off-line methods. Biotechnology Progress 16:866-871 (2000).
Per.C6® is a registered trademark of Crucell (Leiden, The Netherlands).
About the Author
Terrell Johnson, Ph.D., is a Senior Scientist; Teena Rull, M.S., is a Scientist;
Brad Fuhr, B.S., and Brigid DeLong, B.S., are Associate Scientists; and
Mathew Caple is a manager in the Biotechnology R&D Department at Sigma-Aldrich,
St. Louis, MO.
| ORDERING INFORMATION |
| Product Code |
Product Description |
Unit |
| G 9916 |
Gene Therapy Medium-3 for Adenovirus Production |
500 ml
1 liter
6 x 500 ml
6 x 1 liter |
|