HIS-SelectTM Affinity Gels and Plates for Affinity
Capture of Histidine-Containing Proteins
By Ned Watson, Patricia Lindbloom*, Kelly Foster, Jodi Zobrist, Michael
Scott, Rick Mehigh and John Dapron
Sigma-Aldrich Corporation, St. Louis,
MO and Purdue University*, West Lafayette, IN, USA
Introduction
Affinity-based protein capture methods have become standard techniques
for isolation and purification of proteins.1,2 The development
of epitope tagging systems for recombinant proteins and small scale, affinity-based
molecular pull-down methods, such as immunoprecipitation, have enabled
more rapid and detailed study of expression, modification and interaction
of proteins in a wide variety of biological systems.3,4 One
of the most popular and widely used tagging systems involves relatively
short peptide epitope tags containing histidine residues. The histidine-containing
tags allow easy, one-step purification of the tagged fusion protein by
immobilized metal affinity chromatography (IMAC).
IMAC has been a versatile purification tool since first introduced by
Porath et al. in 1975.5,6 The process is based on the potential
for protein/peptide amino acid side chains to form coordination bonds
with immobilized metals. The amino acid side chains generally will contain
oxygen, nitrogen and/or sulfur. Cysteine and histidine residues are the
primary points of interaction on proteins. It has been demonstrated that
proteins containing clustered histidine residues will interact strongly
and with high selectivity for chelated nickel.6
The HIS-Select nickel chelate is a proprietary tetradentate nitriloacetic
acid (NTA) analog chelate and spacer developed at Sigma-Aldrich. This
chelate provides strong coordination with nickel while still presenting
two nickel coordination sites for efficient complexing with recombinant
fusion proteins containing histidine (Figure 1). In addition, the chelate
contains an integral, neutral spacer providing for coupling to various
matrices by stable uncharged bonding. This chelate chemistry is common
throughout the HIS-Select line of products and ensures high selectivity
and stable, reproducible performance. In addition, the uncharged spacer
results in lower non-specific protein binding than typical IMAC affinity
capture products. This new, high performance chelate is used for all the
new IMAC affinity products discussed here.
Materials and Methods
All materials were supplied by Sigma-Aldrich Corporation (St. Louis, MO),
unless otherwise stated.
HIS-Select HC (High Capacity) Nickel Affinity Gel purification of
a histidine-tagged protein from Escherichia coli
A recombinant clone expressing bacterial alkaline phosphatase protein
(BAP) with an N-terminal histidine-containing tag (HAT tag, Clonetech,
Palo Alto, CA; sequence on p. 10) was grown to early log phase with shaking
at 37 °C in 50 ml of EZMix Terrific Broth (Product Code T9179).
The cells were induced with 1 mM isopropyl b-D-thiogalactopyranoside [IPTG]
(Product Code I6758) and allowed to grow for an additional 5 hours. The
cells were harvested by centrifugation at 4 °C at 5,000 x g. The clear
supernatant was removed and the cell paste was frozen at 20 °C.
About 1.2 g of cell paste of the recombinant Escherichia coli expressing
the N-terminal HAT-tagged BAP (HAT-BAP) was extracted in 6 ml of 50 mM
sodium phosphate, 0.3 M sodium chloride, pH 8.0. The cells were disrupted
by sonication for 2 minutes on ice. The extract was clarified by centrifugation
for 15 minutes at 20,000 x g at 4 °C. The clear supernatant was recovered
and loaded on a 0.5 ml column of HIS-Select HC Nickel Affinity Gel (Product
Code P6611) and allowed to flow through by gravity. The column was washed
with 10 ml of equilibration buffer (50 mM sodium phosphate, 0.3 M sodium
chloride, 10 mM imidazole, pH 8.0). The column was then eluted with 1.5
ml of 50 mM sodium phosphate, 0.3 M sodium chloride, 200 mM imidazole,
pH 8.0. The lysate and column fractions were analyzed on a precast 4-20%
tris-glycine gel (Invitrogen, Carlsbad, CA). The proteins were visualized
by staining for one hour with EZBlue Gel Staining Reagent (Product
Code G1041), followed by destaining overnight with deionized water. Protein
concentrations were determined by the Bradford protein assay (Product
Code B6916).
Preparation of mammalian cell lysates
COS-7 cells were grown in Dulbeccos Modified Eagles Medium
(DMEM; Product Code D6171) containing 10% Fetal Bovine Serum (FBS; Product
Code F2442) in 100 mm diameter tissue culture plates (Product Code C6546). When the cells were confluent (107 cells/plate), the plates were
set on ice, the medium was removed, and the adherent cells were washed
twice with 10 ml of ice cold Dulbeccos phosphate buffered saline
(Product Code D8537). The cells were lysed directly by adding 1 ml ice
cold RIPA lysis buffer [150 mM NaCl, 1.0% Igepal CA-630 (NP-40; Product
Code I3021), 0.5% sodium deoxycholate (Product Code D6750), 0.1% sodium
dodecyl sulfate (SDS; Product Code L6026), 50 mM Tris, pH 8.0] to each
plate and scraping rapidly with a cell scraper. Mammalian Protease Inhibitor
Cocktail (100 µl; Product Code P8340) was added to each lysate
and the lysates were used immediately or quick-frozen in liquid nitrogen
and stored at 70 °C until needed.
Small-scale affinity capture with EZview Red HIS-Select HC
Nickel Affinity Gel
COS-7 lysates were clarified by centrifugation in a microcentrifuge for
10 minutes at 8,000 x g at 4 °C. The clear supernatants were recovered
and, subsequently, purified HAT-BAP protein (5 µg/ml lysate) was
added in varying amounts. The resulting spiked lysates were incubated
with 50 µl packed gel volume of standard HIS-Select HC Nickel Affinity
Gel or EZview Red HIS-Select HC Nickel Affinity Gel (pre-washed and equilibrated
in RIPA buffer; Product Code E3528) for 1 hour with mixing at 4 °C.
The affinity beads were collected by centrifugation for 30 seconds at
8,000 x g and the supernatants were removed by aspiration. The pellets
were washed three times with 1 ml of RIPA buffer per wash and collected
by centrifugation as above. After aspirating the final wash supernatants,
the affinity bead pellets were each suspended in 25 µl of RIPA buffer
and 25 µl of 2X Laemmli sample buffer (Product Code S3401) and
analyzed by denaturing SDS-polyacrylamide gel electrophoresis (SDS-PAGE)
and immunoblotting as indicated.
Affinity capture with HIS-Select HS (High Sensitivity) Nickel-Coated
96-well plate
One gram of Escherichia coli cell paste was suspended in 10 ml 50 mM sodium
phosphate, 0.3 M sodium chloride, pH 8.0, and lysed by sonication. The
lysate was clarified by centrifugation at 10,000 x g at 4 °C and the
clear supernatant was collected. Purified HAT-BAP fusion protein was spiked
into the lysate supernatant at various concentrations as indicated. A
volume of 200 µl of spiked lysate containing a total of 0.25 ng
to 32 ng HAT-BAP was added per well in quadruplicate wells of a HIS-Select
HS Nickel-Coated 96-well Plate or competitors plate (Product Code
S5688) and incubated for 3 hours at room temperature. The liquid was
removed from the wells by aspiration and each well was washed three times
with 300 µl Phosphate Buffered Saline containing 0.05% Tween 20
(PBST; Product Code P3563) to remove unbound material. After aspirating
the final wash, the amount of HAT-BAP protein captured per well was detected
directly by an enzyme-linked immunoassay. Each ELISA well was incubated
for 45 minutes at room temperature with a 1:10,000 dilution of anti-BAP
monoclonal antibody (Product Code B6804). Subsequently, each well was
washed three times with 300 µl PBST. After the last wash was removed,
each well was incubated for 45 minutes at room temperature with anti-Mouse
IgG HRP conjugate (Product Code A4416). The secondary antibody solution
was removed and each well was washed four times as above. Each well was
then assayed by incubating with 200 µl of TMB substrate (Product
Code T8665) for 40 minutes at room temperature. The reactions were stopped
by adding 100 ml of 1 M sulfuric acid per well. After mixing, the wells
were read at A450 and each set of quadruplicate well values was averaged
and corrected for background absorbance in negative control wells.
Affinity capture with HIS-Select HC Nickel Coated 96-well plate
For binding capacity comparisons, an excess of purified HAT-BAP (30 µg)
in 200 ml TBS was incubated in triplicate wells of a HIS-Select HC Nickel
Coated 96-well Plate (Product Code S5563) or competitor IMAC affinity
plates for 4 hours at room temperature. The samples were removed from
the wells by aspiration and each well was washed three times with 300
ml Phosphate Buffered Saline containing 0.05% Tween 20 and one time with
deionized water to remove residual detergent. After aspirating the final
wash, the total protein captured per well was determined directly in the
wells by Bicinchoninic Acid protein assay (Product Code BCA-1) and the
triplicate well values were averaged.
Results and Discussion
IMAC has become a popular and powerful means of rapidly purifying recombinant
proteins with short poly amino acid tags containing histidine residues.
Many current IMAC affinity capture matrices contain positively charged
spacer arms that attach the metal chelate affinity group to the support
matrix. Although such IMAC affinity capture matrices are widely used,
problems with non-specific protein binding can occur due to the nature
of the matrix and the charge on the spacer arm. Therefore, we developed
a novel, proprietary tetradentate nitriloacetic acid (NTA) analog chelate
with an uncharged spacer arm in order to produce high performance IMAC
affinity matrices, which are efficient for specifically capturing histidine-tagged
proteins with low non-specific protein binding. This improved nickel chelate
was used to make both resin and plate-based capture systems for IMAC purification
of histidine-tagged recombinant proteins.
We developed a high capacity agarose-based nickel chelate affinity resin,
HIS-Select HC Nickel Affinity Gel, and demonstrated its performance for
IMAC purification of a HAT-tagged recombinant protein from Escherichia
coli. The HAT tag is a 19 amino acid tag, which contains 6 histidine residues
interspersed with other amino acids. It is originally derived from a natural
protein sequence for chicken lactate dehydrogenase that was known to bind
a metal chelate. Recombinant HAT-BAP was expressed in Escherichia coli
and purified in one step on a small HIS-Select HC Nickel Affinity Gel
column. The purified, bound HAT-BAP protein was eluted in the presence
of 200 mM imidazole (Figure 2) . Analysis of the column fractionation indicated that
most of the HAT-BAP protein from the lysate bound the column (compare Figure 2, lanes 2 and 3). The purified, eluted HAT-BAP protein
was greater than 95% pure after this one-step purification with little
non-specific protein binding observed (Figure 2, lane 4).
Often small-scale affinity capture methods, such as immunoprecipitation,
are used to analyze protein expression, post-translational modifications,
or protein-protein interactions. A major disadvantage of these small scale
affinity capture procedures, as commonly practiced, is that the affinity
matrix is difficult to see in microcentrifuge tubes that are used for
the complex formation, purification and wash steps. This difficulty in
visualization leads to inefficient and tedious manipulations, and often
results in loss of material and quantitative variability of results. Therefore,
we developed unique, highly visible red colored HIS-Select affinity beads,
EZview Red HIS-Select Affinity Gel (Product Code E3528), to be
used in small-scale affinity capture of histidine-
tagged target proteins.
To compare EZview Red HIS-Select HC Nickel Affinity Gel with standard
HIS-Select HC Nickel Affinity Gel for recovery of the HAT-BAP protein
from mammalian cell lysates, we spiked purified HAT-BAP into COS-7 cell
lysates and performed small-scale affinity capture with both affinity
resins. After binding and washing steps, the captured target protein was
eluted from the affinity beads subjected to SDS-PAGE and analyzed by immunoblotting.
The increased visibility of the EZview resin made washing and supernatant
removal manipulations faster and easier during the experiment than with
the standard HIS-Select resin. Immunostaining of the Western blots revealed
that a similar amount of HAT-BAP protein was recovered from COS-7 lysates
with both the standard HIS-Select and the EZview Red HIS-Select HC Nickel
Affinity Gel (Figure 3, compare lanes 4 and 5). In addition, no differences in
non-specific background protein binding were observed (Figure 3, compare lanes 2 and 3).
For parallel processing of numerous samples or for other high-throughput
applications such as high-throughput protein capture or screening assays,
we employed the same unique nickel chelate to make multi-well HIS-Select
capture plates. We developed a high sensitivity plate,
HIS-Select HS Nickel 96-well plate (Product Code S5688), for capture
and low level detection of histidine-tagged proteins. The performance
of this high sensitivity IMAC plate was compared to a competitors
IMAC affinity plate for the capture of a histidine-tagged target protein,
HAT-BAP, from a cell lysate. The HIS-Select HS Nickel 96-well plate captured
and allowed detection of spiked HAT-BAP protein from the lysate over a
concentration range of 1 to 32 ng/well, as measured in a standard enzyme-linked
immunoassay (Figure 4).
No protein was detected in this range in the same assay using a competitors
IMAC plate. The lower limit of detection for the HIS-Select HS Nickel
96-well plate was less than 1 ng/well using this standard enzyme-linked
assay. Therefore, this plate is an excellent platform for capture and
detection of low levels of recombinant fusion proteins with histidine-containing
tags.
In addition, novel HIS-Select HC Nickel multiwell plates (96-well plate,
Product Code S5563; 384-well plate, Product Code H1661) were developed,
which have a high capacity for rapid, simultaneous IMAC purification of
histidine-containing proteins. The wells of these plates are coated with
a proprietary high-density, large pore size, nickel chelate matrix, which
provides a much higher capacity for binding proteins with histidine-containing
tags. This allows rapid affinity purification of histidine-containing
proteins from numerous samples in sufficient quantity per well (typically
> 4 µg per well) for analysis by standard biochemical methods.
These methods include direct quantitation of captured protein by standard
protein assays, such as Bradford or BCA assays, and rapid analysis of
the captured histidine-containing protein, or protein complexes, by SDS-PAGE,
Western immunoblotting and/or mass spectrometry. The binding capacity
of the HIS-Select HC Nickel 96-well plates was compared to plates of competitors
using an excess of purified HAT-BAP protein. Even though the HAT tag is
a relatively weak nickel-binding tag, nearly 3 µg HAT-BAP was bound
per well for the HIS-Select HC Nickel 96-well plate (Figure 5). The amount of HAT-BAP bound to the wells of the HIS-Select
HC Nickel 96-well plate was much greater than the amount bound to wells
of the competitor plates, which was at the lower limit of detection. These
high capacity IMAC affinity plates are extremely useful for isolation
and characterization of histidine-tagged proteins and for parallel purification
of large numbers of histidine-tagged proteins for applications such as
biochemical and structure/function analysis of multiple deletion and point
mutant protein variants.
Conclusions
We developed a unique, stable tetradentate nickel chelate that utilizes
a non-charged hydrophilic spacer to reduce non- specific protein binding.
We have attached this proprietary chelate to both resin and multi-well
plates to provide improved platforms for the purification and analysis
of proteins containing histidine tags. The new IMAC affinity matrices
are a significant improvement over previous IMAC affinity resins and plates
and should be more useful for the capture and analysis of histidine-tagged
proteins for a number of different applications in the future.
Acknowledgements
The authors would like to thank Dr. Bill Kappel of the Protein R&D group
at Sigma-Aldrich for helpful discussions and critical reading of the manuscript.
References
1. Scouten, W. H., Affinity Chromatography: Bioselective Absorption on
Inert Matrices. John Wiley and Sons, Inc., New York (1981).
2. Street, G. (Ed.), Highly Selective Separations in Biotechnology. Chapman
and Hall, London (1994).
3. Kolodziej, P. A. and Young, R. A., Epitope tagging and protein surveillance.
Methods Enzymol. 194, 508-519 (1991).
4. Harlow, E. and Lane, D., Antibodies: A Laboratory Manual. Cold Spring
Harbor Laboratory Press, New York, p. 423-470 (1988).
5. Porath, J., et al., Metal chelate affinity chromatography, a new approach
to protein fractionation. Nature, 258, 598-599 (1975).
6. Sulkowski, E., Purification of proteins by IMAC. Trends in Biotechnology,
3, 1-12 (1985).
About the Authors
Ned Watson, Ph.D. and Kelly Foster, B.S. are Senior Scientists; Jodi Zobrist,
M.S. and Michael D. Scott, M.S. are Scientists; and Rick Mehigh, Ph.D.
and John Dapron, B.S. are Principal Scientists in the Biotechnology R&D
Department at Sigma-Aldrich, St. Louis, MO. Patricia Lindbloom is a Sigma
Undergraduate Co-op student from the Biology Department at Purdue University,
West Lafayette, IN.
| ORDERING INFORMATION |
| Product Code |
Product Description |
Unit |
| P 6611 |
HIS-Select HC Nickel Affinity Gel* |
5 ml
25 ml |
| E 3528 |
EZview Red HIS-Select HC Nickel Affinity Gel* |
1 ml
5 x 1 ml |
| S 5688 |
HIS-Select HS Nickel 96-Well Strip Plate* |
1 plate
5 plates |
| S 5563 |
HIS-Select HC Nickel 96-Well Plate* |
1 plate
5 plates |
| H 1661 |
HIS-Select HC Nickel 384-Well Plate* |
1 plate
5 plates |
*Patent Pending
Amino acid sequence of the HAT tag
Lys-Asp-His-Leu-Ile-His-Asn-Val-His-Lys-Glu-Glu-His-Ala-His-Ala-His-Asn-Lys