Leptomycin B: A Powerful Antibiotic Tool for Studying Nuclear Transport
By Ned Watson
Sigma-Aldrich Corporation, St. Louis, MO, USA
Introduction
Leptomycin B (LMB) is an antibiotic with anti-fungal and anti-tumor activity that
was first discovered and purified from the fermentation broth and mycelia
of Streptomyces.1-4 LMB (C33H48O6,MW
540) is an unsaturated, branched chain fatty acid with a terminal lactone
ring (Figure
1).2 Recently, this antibiotic has become an important
tool for studying nuclear localization and trafficking in eukaryotic cells,
due to specific inhibition of the CRM1/exportin1 nuclear export pathway.
Discussion
Proteins
and other macromolecules constantly move into and out of eukaryotic cell
nuclei. Bi-directional nuclear transport is a regulated, signal-mediated
process that occurs through specific proteinaceous structures, the nuclear
pore complexes, which span the nuclear envelope.5-8 In the
last decade, there has been much progress in understanding the composition
and structure of the nuclear pore complex and the mechanisms of nuclear
transport. A number of important discoveries have helped elucidate the
protein import process. This includes characterization of the classic
nuclear localization signal (NLS), discovery of NLS recognition by the
soluble adaptor protein, importin- ;
, the role of the importin-
receptor subunit for import and nuclear release of cargo protein, and
the role of Ran-GTP in cargo discharge and transport directionality.7-10
The importin-
protein is one member of a conserved family of transport receptors, also
termed karyopherins. Each member recognizes a distinct NLS. Karyopherins
can also bind the small GTP-binding protein, Ran, and nuclear pore complex
proteins (nucleoporins).
More
recently, details of the related mechanism for nuclear export have emerged.7-10
Much of the early progress has come from studies of the HIV Rev protein
and the cellular PKI protein (protein kinase A inhibitor). These studies
led to the discovery and characterization of the leucine-rich nuclear
export signal (NES) and the CRM1/exportin1 pathway of nuclear export.
CRM1 is a karyopherin specific for nuclear export (exportin). A number
of reports have now established that the CRM1 protein directly binds proteins
that contain a leucine-rich NES.11-14 In addition, the export
mechanism appears to involve CRM1 binding to both Ran-GTP and nucleoporins.15
The
stability of the karyopherin/cargo protein interaction is modulated by
the nucleotide-bound state of Ran, which modulates import and export differentially.9,10
Interactions of importins with their cargo proteins are disrupted by Ran-GTP,
which is maintained at high levels in the nucleus by the presence of its
guanine nucleotide exchange factor, Ran-GEF, specifically localized in
the nuclear compartment. This results in the rapid discharge of the cargo
when the complex reaches the nucleus. In contrast, the interactions of
exportins with their respective cargo proteins are stabilized by Ran-GTP
binding. These interactions are disrupted upon Ran-GTP hydrolysis, which
occurs rapidly in the cytoplasm, due to the presence of high levels of
the Ran GTPase activating protein, Ran-GAP, in the cytoplasmic compartment.
Thus, the compartment-specific localization of the regulators of the Ran
nucleotide-bound state is responsible for the Ran-GTP gradient across
the nuclear membrane. The Ran-GTP gradient effectively determines the
directionality of nuclear transport by regulating cargo binding and release.
Tools
available for perturbing nuclear transport are still somewhat limited
and include genetic loss of function mutants or dominant mutants in karyopherins
or the Ran GTPase regulation pathway. While there are no specific drugs
for inhibiting nuclear import pathways, there is a drug that inhibits
nuclear export. It has recently been shown that the anti-fungal antibiotic,
Leptomycin B, specifically and potently inhibits the CRM1/exportin 1 pathway
of nuclear export by directly binding the CRM1 protein.
Leptomycin
B has been an important tool in the elucidation of the role of CRM1/exportin
1 in the export process. The inhibition of growth of fission yeast and
mammalian cells by LMB4 was shown to be due to the inhibitory
effect of LMB on CRM1-mediated processes.16 Later studies on
HIV-Rev protein export determined that LMB is an inhibitor of nuclear
export.17 It now has been demonstrated clearly that the cellular
effects of LMB are due to inhibition of nuclear export as a consequence
of LMB binding covalently to the CRM1 protein. Binding of LMB to CMR1
occurs in its conserved central region at a critical cysteine residue
and prevents formation of the complex between CRM1 and the NES of cargo
proteins.18,19 Unlike fission yeast and mammalian cells, Saccharomyces
cerevisiae does not show toxic effects of LMB treatment. However,
when a single threonine in the conserved central region of the S. cerevisiae
CRM1 protein (Thr539) is changed to cysteine, the resulting strain becomes
fully sensitive to LMB.20 Thus, CRM1 structure and function
appears to be conserved throughout eukaryotes.
It
is becoming increasingly clear to cell biologists that the steady state
localization of proteins does not always reflect their biologically important,
functional sites of action. For example, steady state localization of
a protein in the cytoplasm may not allow its dynamics in and out of the
nucleus to be detected. However, the use of Leptomycin B as a tool to
block CRM1-mediated nuclear export results in the accumulation of NES-containing
proteins in the nucleus and allows their detection in the nuclear compartment
(Figure
2).
Summary
Understanding
the mode of action for Leptomycin B inhibition of the CRM1/exportin 1
nuclear export pathway has significantly increased the utility of LMB
for cell biology studies on nuclear transport and localization. Recently,
Leptomycin B has been used in many systems to demonstrate nuclear localization
and CRM1 pathway-dependent export of a number of proteins, including actin,
transcription factors, kinases and cell cycle regulators (Figure
3).21-27 The creation of a LMB sensitive strain of S.
cerevisiae has further increased the utility of this antibiotic.20
Usage of Leptomycin B for study of nuclear localization and export should
continue to increase in the future, as more purified and reliable supplies
become commonly available to researchers.
Acknowledgements
The
author would like to thank Dr. Susan Wente, Dept. Cell Biology and Physiology,
Washington University School of Medicine, for helpful discussions and
for critical reading of the manuscript, and Tom Rutkoski, Sigma Undergraduate
Co-op Program student, Pennsylvania State University, for help with Figure
2. I am also very grateful to Dr. Helen Pwinica-Worms, Howard Hughes
Medical Institute and Dept. Cell Biology and Physiology, Washington University
School of Medicine, for kindly providing the images of cells shown in
Figure
3.
References
1.
Hamamoto, T., et al., Leptomycins A and B, new antifungal antibiotics.
I. Taxonomy of the producing strain and their fermentation, purification
and characterization. J. Antibiot. (Tokyo), 36, 639-645 (1983).
2. Hamamoto, T., et al., Leptomycins A and B, new antifungal antibiotics.
II. Structural elucidation. J. Antibiot. (Tokyo), 36, 646-650 (1983).
3. Hamamoto, T., et al., Leptomycins A and B, new antifungal antibiotics.
III. Mode of action on Schizosaccharomyces pombe. J. Antibiot. (Tokyo),
38, 1573-1580 (1985).
4. Yoshida, M., et al., Effects of Leptomycin B on the cell cycle of fibroblasts
and fission yeast cells. Exp. Cell Res. 187, 150-156 (1990).
5. Wente, S. R., Gatekeepers of the nucleus. Science, 288, 1374-1377
(2000).
6. Ryan, K., and Wente, S. R., The nuclear pore complex: a protein machine
bridging the nucleus and cytoplasm. Curr. Opin. Cell Biol., 12,
361-371 (2000).
7. Mattaj, W., and Englmeier, L., Nucleocytoplasmic transport: the soluble
phase. Annu. Rev. Biochem., 67, 265-306 (1998).
8. Gorlich, D., Transport into and out of the cell nucleus. EMBO J., 17,
2721-2727 (1998).
9. Nakielny, S., and Dreyfuss, G., Transport of proteins and RNAs in and
out of the nucleus. Cell, 99, 677-690 (1999).
10. Azuma, Y., and Dasso, M., The role of Ran in nuclear functions. Curr.
Opin. Cell Biol., 12, 302-307 (2000).
11. Stade, K., et al., Exportin 1 (Crm1p) is an essential nuclear export
factor. Cell, 90, 1041-1050 (1997).
12. Fornerod, M., et al., CRM1 is an export receptor for leucine-rich
nuclear export signals. Cell, 90, 1051-1060 (1997).
13. Ossareh-Nazari, B., et al., Evidence for a role of CRM1 in signal-mediated
nuclear protein export. Science, 278, 141-144 (1997).
14. Fukuda, M., et al., CRM1 is responsible for intracellular transport mediated
by the nuclear export signal. Nature, 390, 308-311 (1997).
15. Floer, M., and Blobel, G., Putative reaction intermediates in Crm1
mediated nuclear protein export. J. Biol. Chem., 274, 16279-16286
(1999).
16. Nishi, K., et al., Leptomycin B targets a regulatory cascade of crm1,
a fission yeast nuclear protein, involved in control of higher order chromosome
structure and gene expression. J. Biol. Chem., 269, 6320-6324 (1994).
17. Wolff, B., et al., Leptomycin B is an inhibitor of nuclear export:
inhibition of nucleo-cytoplasmic translocation of the human immunodeficiency
virus (HIV-1) Rev protein and Rev-dependent RNA. Chem. Biol., 4,
139-147 (1997).
18. Kudo, N., et al., Leptomycin B inhibition of signal-mediated nuclear
export by direct binding to CRM1. Exp. Cell Res., 242, 540-547
(1998).
19. Kudo, N., et al., Leptomycin B inactivates CRM1/exportin 1 by covalent
modification at a cysteine residue in the central conserved region. Proc.
Natl. Acad. Sci. USA, 96, 9112-9117 (1999).
20. Neville, M., and Rosbash, M., The NES-Crm1p export pathway is not
a major mRNA export route in Saccharomyces cerevisiae. EMBO J., 18,
3746-3756 (1999).
21. Wada, A., et al., Nuclear export of actin: a novel mechanism regulating
the subcellular localization of a major cytoskeletal protein. EMBO J.,
17, 1635-1641 (1998).
22.
Sachdev, S., and Hannink, M., Loss of Ikßa-mediated control over
nuclear import and DNA binding enables oncogenic activation of c-Rel.
Mol. Cell. Biol., 18, 5445-5456 (1998).
23. Kudo, N., et al., A novel nuclear export signal sensitive to oxidative
stress in the fission yeast transcription factor Pap1. J. Biol. Chem.,
274, 15151-15158 (1999).
24. Huang, T. T., et al., A nuclear export signal in the N-terminal regulatory
domain of Ikßa controls cytoplasmic localization of inactive NF-kß/IkBa
complexes. Proc. Natl. Acad. Sci. USA, 97, 1014-1019 (2000).
25. Fukada, M., et al., Interaction of MAP kinase with MAP kinase kinase:
its possible role in control of nucleocytoplasmic transport of MAP kinase.
EMBO J., 16, 1901-1908 (1997).
26. Engel, K., et al., Leptomycin B-sensitive nuclear export of MAPKAP
kinase 2 is regulated by phosphorylation. EMBO J., 17, 3363-3371
(1998).
27. Taagepera, S., et al., Nuclear-cytoplasmic shuttling of C-ABL tyrosine
kinase. Proc. Natl. Acad. Sci. USA, 95, 7457-7462 (1999).
| ORDERING INFORMATION |
| Product Code |
Product Name |
Unit |
|
L2913
|
Leptomycin B
> 95% by HPLC
0.5 µg in 100 µl 70% methanol
|
0.5 µg