by Ned Watson
Sigma-Aldrich Corporation, St. Louis, MO, USA
Introduction
Fractionation of cells into their subcellular components has long been
a central approach in cell biology. Subcellular fractionation techniques
have been used widely to study structure and function of organelles and
subcellular compartments, as well as to examine the location, processing,
and trafficking of molecular components.1 The object of most
subcellular fractionation procedures is to obtain cellular organelles
and macromolecules in a functional state, in which they retain most of
their original biochemical properties. This is usually achieved by employing
cell lysis by gentle mechanical means or with mild detergents, followed
by fractionation of cellular components by differential centrifugation.2,3
The study of the cell nucleus and nuclear events has been necessary
for understanding a number of processes of primary importance in cell
biology, including chromatin structure, transcriptional regulation of
gene expression, RNA synthesis and processing, mechanism and regulation
of bi-directional nuclear transport, and nuclear apoptosis. Since preparation
of nuclei or nuclear extracts is often the first step in studying nuclear
components and events, a number of different techniques for isolating
nuclei have been described. These methods vary considerably, depending
on species or tissue type and the downstream application for the isolated
nuclei. This review describes the most widely used methods for isolation
and use of mammalian cell nuclei, and introduces several new products
that incorporate improvements to standard methods.
Discussion
It is important to choose carefully a method of nuclei isolation that
will yield nuclei with the desired properties. In some cases, such as
investigations of nuclear transport and in vitro studies of nuclei
assembly and disassembly processes, it is necessary to isolate nuclei
with intact nuclear envelopes.4 In these cases the isolated
nuclei have intact inner and outer membranes and nuclear pore structures.
Since the outer nuclear membrane is continuous with the rough endoplasmic
reticulum (RER) of the cytoplasm, these nuclei can have significant cytoplasmic
contamination that can interfere with purification of nuclear components
or obscure the proper interpretation of molecular localization studies.
In addition, the nuclear membrane is a selective barrier, which can compromise
rapid and efficient entry of small molecules, such as nucleotide precursors
used for labeling nascent RNA chains in transcription run-off experiments.
Therefore, procedures which remove the nuclear membranes, but yield otherwise
intact nuclei, are frequently preferred for the study of nucleoplasmic
components and functions other than nuclear transport.
There are several widely accepted standard methods for isolating nuclei
from mammalian cells5-7 that employ gentle, non-ionic detergents
and yield nuclei free from RER and other cytoplasmic contamination. The
resulting nuclei are functional for the synthesis and extension of endogenous
RNA primary transcripts. The method for nuclei isolation from tissue culture
cells utilizes a hypotonic Nonidet P-40 or Igepal CA-360 detergent lysis
buffer.6,7 Since nuclei are the largest organelles in the cell,
they are easily separated from other organelles and detergent-soluble
contaminants by low speed centrifugation and further purified by repeated
washes in the same lysis buffer. For isolation of nuclei from solid tissues
or from cell lines with fragile nuclei, the nuclei are purified by centrifugation
through a dense sucrose cushion to protect nuclei and strip away cytoplasmic
contaminants.5-7
We have recently improved the method for isolation of nuclei from adherent
tissue culture cell lines. In the standard protocol6,7 cells
are harvested by scraping in phosphate buffered saline (PBS) and collected
by centrifugation, before being lysed in the detergent containing lysis
buffer. However, scraping cells in PBS yields a heterogeneous population
of intact cells, physically damaged cells, cell debris, and free nuclei.
In addition, the harvest of adherent cells by scraping in PBS is inefficient.
It often results in low and variable yields of nuclei, and delay in time
before all the cells are lysed. In the improved method (Nuclei EZ Prep
Nuclei Isolation Kit, Product Code: NUC-101), cells are harvested and
lysed simultaneously. This modification is critical for rapid, efficient
and complete lysis of the cells, and decreases the possibility of artifacts
due to the cell harvesting procedure. The isolated nuclei appeared to
be structurally intact and free from RER and cytoplasmic debris (Figure
1).
For nuclei isolation from animal tissues, from cultured cells with fragile
nuclei, or from cultured cells that are difficult to harvest or lyse (e.g.,
epithelial-like cells with tight junctions), the preferred method remains
detergent and physical lysis by homogenization. This is done in an iso-osmotic
sucrose buffer with subsequent purification of the nuclei by ultracentrifugation
through a dense (2 M) sucrose cushion.5-7 However, many mammalian
nuclei are not dense enough to pass through a 2 M sucrose cushion, resulting
in poor yields of purified nuclei. We recently improved the standard protocol
by decreasing the sucrose cushion concentration from 2 M to 1.8 M sucrose
(Nuclei PURE Prep Nuclei Isolation Kit, Product Code: NUC-201). Nuclei
from a variety of mammalian cells passed through the 1.8 M sucrose cushion,
resulting in greatly improved yields. The 1.8 M sucrose cushion purified
nuclei appeared to be structurally intact and free from RER and cytoplasmic
debris (Figure
2).
In addition to high yields of pure nuclei (Figure
1 and 2),
the resulting nuclei from both of the improved methods are also functional
for run-off synthesis of RNA polymerase II dependent RNA transcripts (Figure
3). Therefore, these procedures should be useful for examining the
transcriptional state of mammalian cells in nuclear transcription run-off
experiments.
Also, nuclei can be isolated and nuclear extracts can be prepared in
one integrated protocol (Nu-CLEAR Extraction Kit, Product Code:
N-XTRACT). Such nuclear extracts are useful for analyzing transcription
factors by Electrophoretic Mobility Shift Assays (Mobility Shift Optimization
Kit, Product Code: SHIFT-1) and for in vitro transcription reactions.
All nuclei isolation procedures have some common potential technical
problems. Contamination by endogenous proteases or nucleases, and extraction
or physical perturbations by detergents may cause adverse effects on the
quality of the nuclei. The nuclei may become more fragile due to the loss
of nuclear membranes and, consequently, the purified nuclei may aggregate
if excessive DNA leakage occurs. These potential problems are usually
overcome by rapidly isolating the nuclei from fresh cells or tissue and
by keeping the nuclei cold (4 ºC) during the isolation procedure.
If necessary, protease and/or nuclease inhibitors may be added to the
lysis buffers to minimize enzymatic hydrolysis of the molecules of interest.
Summary
Methods of nuclei isolation will continue to be useful for many common
cell biology applications, such as purification of nuclear components
(chromatin, genomic DNA, histones and nuclear RNA/RNP) and for functional
studies, such as examination of the transcriptional status of cells by
in vitro transcription run-off analysis.5-7 Improved
techniques, such as those reported here, will be increasingly useful for
isolation of nuclei for a variety of applications, including newer methods
such as in vitro nuclear apoptosis assays,8,9 and transcription
profiling. In addition, these and other cellular fractionation methods
will be important in the future for the emerging areas of functional genomics
and proteomics.10
References
1. Graham, J.M., and Rickwood, D., Subcellular Fractionation: A Practical
Approach, (IRL Press, Oxford, 1997).
2. Claude, A., The coming of age of the cell. Science, 189, 433-435
(1975).
3. de Duve, C., and Beaufay, H., A short history of tissue fractionation.
J. Cell Biol., 91, 293s-299s (1981).
4. Kihlmark, M., and Hallberg, E., Preparation of Nuclei and Nuclear Envelopes,
in Cell Biology: A Laboratory Handbook, Vol. 2, Celis, J.E.
(Ed.) pp. 152-158 (Academic Press, San Diego, 1998).
5. Marzluff, W.F., and Huang, R.C.C., Transcription of RNA in Isolated
Nuclei, in Transcription and Translation: A Practical Approach,
Hames, B.D., and Higgens, S.J. (Eds.) pp. 89-129 (IRL Press, Oxford, UK,
1984).
6. Greenberg, M.E., and Bender, T.P., Identification of Newly Transcribed
RNA, in Current Protocols in Molecular Biology, Ausubel, F.M.,
et al., (Eds.) pp. 4.10.1-4.10.11 (John Wiley and Sons, New York, 1997).
7. Farrell, Jr., R.E., Analysis of Nuclear RNA, in RNA Methodologies:
A Laboratory Guide for Isolation and Characterization, Farrell, Jr.,
R.E., (Ed.) pp. 406-437 (Academic Press, San Diego, 1998).
8. Lazebnik, Y.A., et al., Nuclear events of apoptosis in vitro
in cell-free mitotic extracts: a model system for analysis of the active
phase of apoptosis. J. Cell Biol., 123, 7-22 (1993).
9. Juin, P., et al., Induction of a caspase-3-like activity by calcium
in normal cytosolic extracts triggers nuclear apoptosis in a cell-free
system. J. Biol. Chem., 273, 17559-17564 (1998).
10. Patton, W.F., Proteome analysis. II. Protein subcellular redistribution:
linking physiology to genomics via the proteome and separation technologies
involved. J. Chromotography B. Biomed. Sci. Appl., 722, 203-223
(1999).
Reprinted from Neurotransmissions, 15(4), 18-21, (1999), a newsletter
of Sigma-RBI.
Nu-CLEAR is a trademark of the Sigma-Aldrich Corporation.
About the Author
Ned Watson, Ph.D., is a senior scientist in Gene Expression Analysis
R&D at Sigma-Aldrich, St. Louis, MO.
| ORDERING
INFORMATION |
| Product Code |
Product Name |
Unit |
| NUC-101 |
Nuclei EZ Prep
nuclei isolation kit |
1 kit (25 nuclei
preps) |
|
NUC-201 |
Nuclei PURE
Prep nuclei isolation kit |
1 kit (15 nuclei
preps) |
| RELATED
PRODUCTS |
| Product Code |
Product Name |
Unit |
|
N-XTRACT |
Nu-CLEAR
extraction kit |
1 kit (100 preps) |
|
SHIFT-1 |
Mobility Shift
Optimization kit |
1 kit
(100 trials) |
|
P8340
|
Protease
Inhibitor Cocktail
|
1 ml |
| |
|
5 ml |
|
R7253
|
Ribonuclease
Inhibitor from human placenta |
300
units |
| |
|
1500 units |
|
|
5000 units |
|
|
30,000 units |