Molecular Biology
Automated Sequencing
Product Lines
Product Highlights
Key Resources
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SigmaSpin™ Post-Reaction Clean-Up
Automated Sequencing
For removal of unincorporated dyes, excess salts and other interfering components from sequencing reactions.
SigmaSpin™ Post-Reaction Clean-Up Columns
SigmaSpin™ Post-Reaction Clean-Up Columns are ideal for lower throughput applications, such as clean-up of probe labeling reactions or small numbers of sequencing reactions.
These columns can accept sample volumes up to
100 µl. Each column comes with a collection tube to collect the DNA during centrifugation.
SigmaSpin™ 96-Well Post-Reaction Clean-Up Plates
SigmaSpin™ 96-Well Post-Reaction Clean-Up plates provide a fast, simple, and highly efficient method for removing unincorporated dyes, excess salts, and other interfering reaction components (Fig. 1). Each plate is packed with a pre-hydrated size-exclusion resin, equilibrated with molecular biology grade water, and supplied in our unique plate design with long drip directors to minimize contamination between samples. The plate design also includes a snap-cap bottom seal and a foil seal top to ensure that the resin remains hydrated. SigmaSpin™ has been tested in high-throughput genome centers and core facilities with ABI Prism® 3700, 3100, 310 and 377. Each well can accept sample volumes up to 20 µl.
Ideal for removing
  • Dye-terminator nucleotides and primers from sequencing reactions
  • Radiolabeled nucleotides, primers, and fluorescent dyes from nucleic acid probe labeling reactions
SigmaSpin™ Post-Reaction Clean-Up 96-well plates come ready for immediate use.
Features and Benefits
  • Validated with most automated sequencers and dye terminator chemistries (Fig. 2)
  • Pre-qualified size-exclusion resin guarantees optimum performance
  • Unique drip directors prevent cross-contamination between samples during collection
  • Plates are sealed to eliminate leakage or drying during shipping or storage
  • Suitable for use with multi-channel pipettes and automated workstations
Comparison of Sequencing reaction clean-up methods
Panel A: Sequencing reactions were precipitated with 70% ethanol and placed on ice for thirty minutes. DNA pellets were dried and resuspended in TE solution prior to electrophoresis.
Panel B: Sequencing reactions were subjected to post-reaction clean-up with SigmaSpin™ Post-Reaction Clean-Up 96-Well Plates, according to recommended protocol.
Figure 1. Single stranded M13MP18 plasmid was sequenced with a -21M13 forward sequencing primer using ABI BigDye™ Terminator chemistry. Sequencing reactions were resolved on an ABI Prism® 377 XL instrument with a 48 cm gel cassette containing 4.5% AutoPAGE™ Plus acrylamide at 2.88kV for 7 hrs.
SigmaSpin Removes Interfering Reaction Components
Figure 2. Sequencing reaction was purified using a SigmaSpin 96-well plate. pFLAG™ MAC plasmid (Sigma, E 5644) was sequenced using BigDye™ Terminator v 3.0 chemistry (20 µl total reaction volume; BigDye premix diluted 1:1 with SeqSaver™ [Sigma, S 3938]). Data was generated on an ABI Prism® 3700 DNA Analyzer with POP-6™ polymer and 10X CE Buffer (Sigma, B 4930). Phred 20 > 600.
Ordering Information
Product Product Description Quantity
S5059 SigmaSpin™ Post-Reaction Clean-Up Columns
(with collection tubes)
70/pkg
S4309 SigmaSpin™ Post-Reaction Clean-Up Plates* 2 each
S4434 SigmaSpin™ Post-Reaction Clean-Up Plates* 10 each
S4559 SigmaSpin™ Post-Reaction Clean-Up Plates* 50 each
P4736 48-well Wash Plate 50 each
Z37,212-9 96-Well V-bottom Collection Plates 50 each
*Wash and collection plates included in 2- and 10-each package sizes
back to top
back to Automated Sequencing