Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Chemically Competent & Electrocompetent Cells
Cloning & Expression
High Efficiency Competent Cells for your Sensitive Cloning Applications!
Sigma-Aldrich offers a wide range of chemically competent cells as well as Thunderbolt™ GC10™ electrocompetent cells. Our competent cells are offered at the highest transformation efficiencies for sensitive cloning applications. In addition, we offer a variety of strains to meet your specific needs for protein expression and plasmid propagation.
Features and Benefits
  • Convenient Unipack formats for one time use
  • Strains comparable to DH5α™ and DH10B™
  • High efficiencies
  • T1 phage resistance
Cloning Strains
Expression Strains
Competent Cell Selection Table
The Competent Cell Compendium: Tips and Tools for Successful Transformations" Now Available
Download your copy today!
(765 Kb pdf)
or
Launch the eBook today! Create a shortcut to your desktop!
Other Resources:
     Watch the new technical presentation "Everything you Need to Know about Competent Cells!"
     Calculating Transformation Efficiences
     Genotypes, Phenotypes and Markers - Definitions
     E. coli Markers
     Strains of E. coli
Cloning Strains
Features and Benefits
  • F' episome for single strand rescue
  • Strains for cloning methylated DNA
  • Blue/white selection
Product # 
Product Name
Size
G3169
GC5™ Competent Cells, Uni-pack
10 x 50 µl
20 x 50 µl
G3044
GC5™ Competent Cells, Standard Aliquots
5 x 200 µl
G7419
GC5™ Competent Cells, 96-well
1 EA
4 EA
G2919
GC10™ Competent Cells, Uni-Pack
10 x 50 µl
20 x 50 µl
G2794
GC10™ Competent Cells, Standard Aliquots
5 x 200 µl
T7699
Thunderbolt™ GC10™ ElectroCompetent Cells
5 x 80 µl
5 x 100 µl
J3895
JM109 Competent Cells, Uni-pack
10 x 50 µl
H3788
HB101 Competent Cells, Uni-pack
10 x 50 µl
back to top
Expression Strains
Features and Benefits
  • T1 phage resistance
  • Strains for T7 systems
  • Ion and omp T protease deficient
Product # 
Product Name
Size
B2685
BL21-T1R Competent Cells, Uni-pack
10 x 50 µl
B2935
BL21 (DE3)-T1R Competent Cells, Uni-pack
10 x 50 µl
B3310
BL21 (DE3) pLysS-T1R Competent Cells, Uni-pack
10 x 50 µl
B3435
BL21 (DE3) pLysE-T1R Competent Cells, Uni-pack
10 x 50 µl
back to top
Competent Cell Strain Selection Table:
Feature GC5™ GC10™ Thunderbolt™ GC10™ JM109 HB101 BL21 BL21 (DE3) BL21 (DE3) pLysS BL21 (DE3) pLysE
Comparable to DH5a
X
Comparable to NovaBlue and XL1 Blue
X
Comparable to DH10B™
X
X
Strain Background
K12
K12
K12
K12
K12xB
B
B
B
B
Transformation Efficiency (cfu/ug pUC plasmid DNA)
109
109
1010
108
108
106
107
106
106
Cloning Strain
X
X
X
X
X
Expression Strain
X
X
X
X
Blue/White Selection (with appropriate construct)
X
X
X
X
T1 bacteriophage resistant
X
X
X
X
X
X
X
T7 promoter systems
X
X
X
F' episome for single strand rescue
X
Ideal for cloning methylated DNA
X
X
X
lacI q (higher level of lac repressor)
X
GC5™, GC10™, and Thunderbolt™ are trademarks of GeneChoice ®, Inc.
DH5α™ and DH10B™ are trademarks of Invitrogen Corp.
back to top
back to Cloning & Expression