Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 LB Broth & Terrific Broth Liquid Media
Cloning & Expression
New from Sigma-Aldrich . . .
  • MAXimum convenience with sterile packaging
  • MAXimum performance with greater lot to lot consistency
  • MAXimum value in cost and times savings
Pre-sterilized Liquid Bacterial Media
The ultimate in convenience products - Sigma now offers three different varieties of pre-sterilized, read-to-use liquid medias. LB Broth (Luria Broth Miller) is a standard nutrient media commonly used for cloning applications, plasmid purification and protein expression. Our newest offering, 2X YT Medium, can be used with Escherichia coli and M13 bacteriophage. Terrific Broth is a richer media that would be suitable if higher cell densities and higher yields for plasmids or protein purifications are necessary. All three popular medias are now provided in convenient liquid form to save time and money in your research as well as pre-sterilized and ready for immediate use.
TAKE YOUR LAB TO THE MAX . . .
Product #  Product Name Unit
L2542 LB Broth 500 mL
Y1003 2X YT Medium 500 mL
T5574 Terrific Broth 500 mL
back to Microbial Media & Supplements
back to Cloning & Expression