Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 RapidTransit™ Transformation Kit
Cloning and Expression
Prepare Competent Cells with Speed and Efficiency!
The RapidTransit™ Transformation Kit (R2653) provides an economical and streamlined method to prepare (Figure 1) chemically competent Escherichia coli cells (Figure 2) at efficiencies suitable for standard cloning. RapidTransit reagents eliminate complex buffer preparation and lengthy incubation periods, while reducing the number of procedural steps.
Features and Benefits
  • Fast, economical preparation of competent cells
  • Overcome costs and strain limitations of commercially available cells
  • Suitable for a variety of E. coli strains
  • Eliminate complex buffer preparation
  • Efficiencies suitable for cloning
This kit has been used successfully to prepare competent cells from a wide variety of standard and specialized E. coli strains (Figure 2), including popular K12 cloning strains (GC5™, GC10™, JM109, HB101, DH5a™, and NovaBlue™) and B strains (the BL21 family) used for recombinant protein expression.
The RapidTransit Transformation Kit offers two simple procedures (Figure 1) to prepare competent cells; the Direct Addition Method and the Single-Centrifugation Method. The Direct Addition Method allows for preparation of competent cells, transformation, and recovery within a single tube or well. The Direct Addition Method provides transformation efficiencies of >1 X 105 colony forming units (cfu)/µg supercoiled pUC18 plasmid DNA, whereas the Single-Centrifugation Method provides reproducible transformation efficiencies of >1 X 106 cfu/µg supercoiled pUC18 plasmid DNA, when using strain JM109.
Kit Components
2x RapidTransit Transformation Buffer, 2.5 ml
Recovery Medium, 50 ml
Figure 1. Protocols for preparation of competent E. coli using the RapidTransit Transformation Kit
Figure 2. Comparison of Transformation Efficiency Using Different E. coli Strains.
Competent cells were prepared using the Direct Addition method, for the strains shown above. Cells were transformed with one nanogram pUC18. Error bars indicate standard deviation calculated for the mean of triplicate reactions. A similar pattern of efficiency is observed for cells prepared by Single-Centrifugation method, though approximately 10X higher transformation efficiencies were obtained (data not shown). Transformation efficiency is expressed as colony forming units per µg plasmid.
Product #  Product Name Size
R2653 RapidTransit™ Transformation Kit 50 rxn
back to top
back to Cloning & Expression