Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 S-Gal™
Cloning & Expression
S-Gal™: The Clear Advantage
  • Superior to X-gal for clonal selection
  • Faster, sharper colony color development
  • Temperature stable, can be autoclaved or microwaved
  • Safe, water-soluble reagent
  • Pre-mixed medias provide convenience and time savings
S-Gal is a safe, fast and easy way to differentiate between lac+ and lac- colonies. This non-toxic water-soluble reagent can be added to the media prior to autoclaving. Color determination of black S-Gal colonies begins in as little as 14 hours with sharper contrast for easier colony determination. S-Gal is not light sensitive for convenient storage and usage. The advantages are clear...S-Gal is a superior dye for clonal selection.
Product # 
Product Name
Size
S9811
S-Gal base reagent
100 mg
1 g
5 g
S7313
S-Gal, Sodium Salt
100 mg
1 g
5 g
C4478
S-Gal / LB Agar Blend
500 ml
6 x 500 ml
S1813
S-Gal / Kanamycin / LB Agar Blend
500 ml
6 x 500 ml
S9938
S-Gal / LB Agar Blend (without IPTG)
500 ml
6 x 500 ml
Click here for more information on S-Gal™
back to Microbial Media & Supplements
back to Cloning & Expression