Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SBP
Cloning & Expression
New SBP Bacterial System Expression Vectors
for Simple Purification and Detection Using Streptavidin Products
The new SBP (Streptavidin Binding Peptide) tag provides for one-step purification and detection of recombinant protein from bacterial lysates due to nanomolar affinity for streptavidin. SBP offers high-affinity, high yield, mild elution conditions and simplicity in recombinant protein purification. In addition, the tag provides versatility in downstream detection and assays due to the widely available streptavidin-conjugated fluorescent and enzymatic systems.
  • 38 amino acid tag (MDEKTTGWRGGHVVEGLAGELEQLRARLEHHPQGQREP)
  • One-step purification and detection
  • For use with widely available streptavidin products (resins, enzyme conjugates, plates, etc.)
  • Mild, specific elution of native protein using biotin
  • Higher purity than Maltose Binding Protein or HIS-tag1
  • Expression vectors containing the tac or T7 promoters
Bacterial Expression-tac Promoter
  • Cytoplasmic expression of C-terminal fusion
  • Ampicillin resistance, T1T2 transcriptional terminator, pMB1(derivative of pBR322) origin of replication, and lacI gene for repression of the tac promoter
  • Expression levels using the tac promoter may be in excess of 10 mg/L of culture when using IPTG as a de-repressor
  • Expression in any established Escherichia coli expression host
Product MCS Region
P3996, pTAC-SBP-1
C-terminal SBP
pTAC-SBP-1
back to top
Bacterial Expression-T7 Promoter
  • Cytoplasmic expression
  • N- or C- terminal fusions
  • Ampicillin resistance, T1T2 transcriptional terminator, pMB1 (derivative of pBR322) origin of replication, and lacI gene for repression of the T7 promoter
  • Higher yields of recombinant protein than tac system
  • lac operator (lacO) sequences reduce "leaky" expression
  • Requires hosts with a source of the T7 polymerase such as (DE3) lysogenic strains
  • Enterokinase cleavage of N-terminal SBP
Product MCS Region
P3746, pT7-SBP-1
C-terminal SBP
pT7-SBP-1
E9655, pT7-SBP-2
N-terminal SBP
pT7-SBP-2
Reference
  1. Keefe, A. D., et al., Protein Expr. Purif., 23, 440-446 (2001).
back to top
back to Expression Vectors