Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 BioPORTER® QuikEase™ Protein Delivery Kit
Cloning & Expression
Product No. BPQ24 suitable for 24 reactions
Product No. BPQ96 suitable for 96 reactions
When you need to deliver biologically active proteins into living cells quickly and effectively, at a reduced cost — BioPORTER Protein Delivery Kit is the ideal option.
The BioPORTER reagent reacts quickly and interacts non-covalently with the protein, peptide or other molecules, creating a protective vehicle for immediate delivery into cells. Various molecules (fluorescent-antibody, high and low molecular weight dextran sulfate, caspase 3 and 8, phycoerythrin-BSA, and others) have been successfully delivered into the cytoplasm of a variety of adherent and suspension cells with the BioPORTER reagent. Furthermore, apoptotic proteins like granzyme B, caspase 3 and caspase 8 that were delivered into cells remained functional and induced apoptosis in cultured cells.
BioPORTER is a valuable tool for a variety of studies, including:
  • Intercellular signaling
  • Cell cycle regulation
  • Apoptosis control
  • Oncogenesis
  • Transcripton regulation
BioPorter Kit
Features & Benefits
  • Easy to Use
  • More economical than microinjections or electroporation
  • Reaches optimum levels after four hours of incubation
  • Successfully delivers various molecules such as; fluorescent antibody, high and low molecular weight dextran sulfate, caspase 3 and 8, phycoerythrin-BSA, and others.
  • Delivers effectively into a variety of adherent and suspension cells
  • Apoptopic proteins delivered with BioPORTER have been found to remain functional and induce apoptosis in cultured cells.
back to top
Cell Signaling Homepage